Skip to Content
Merck
All Photos(1)

Documents

702854

Sigma-Aldrich

Copper(II) phthalocyanine

triple-sublimed grade, >99.95% trace metals basis

Synonym(s):

CuPc, Phthalocyanine blue, Pigment Blue 15

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H16CuN8
CAS Number:
Molecular Weight:
576.07
Colour Index Number:
74160
Beilstein:
4121848
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

>99.95% trace metals basis

form

powder

quality

triple-sublimed grade

mp

350 °C

λmax

678 nm

fluorescence

λem 404 nm

Orbital energy

HOMO 5.2 eV 
LUMO 3.5 eV 

OLED Device Performance

ITO/CuPc/NPD/Alq3/C60/Mg:Ag

  • Color: green
  • Max. Luminance: 17200 Cd/m2

ITO/CuPc/NPD/Alq3/LiF/Al
  • Color: red
  • Max. Luminance: 2000 Cd/m2
  • Turn-On Voltage: 7.5 V

ITO/CuPc/NPD/CBP:Firpic (6%)/BAlq3/LiF/Al
  • Color: blue
  • Max. EQE: 4.5 %

OPV Device Performance

ITO/CuPc/PTCDA/In

  • Short-circuit current density (Jsc): 53.1 mA/cm2
  • Open-circuit voltage (Voc): 0.57 V
  • Fill Factor (FF): 0.61
  • Power Conversion Efficiency (PCE): 4.2 %

ITO/PEDOT:PSS/CuPc/C60/BCP/Al
  • Short-circuit current density (Jsc): 18.8 mA/cm2
  • Open-circuit voltage (Voc): 0.58 V
  • Fill Factor (FF): 0.52
  • Power Conversion Efficiency (PCE): 3.6 %

SMILES string

c1ccc2c(c1)C3=NC4=[N@@H]5C(=Nc6n7c(N=C8c9ccccc9C%10=[N@@H]8[Cu]57N3C2=N%10)c%11ccccc6%11)c%12ccccc4%12

InChI

1S/C32H16N8.Cu/c1-2-10-18-17(9-1)25-33-26(18)38-28-21-13-5-6-14-22(21)30(35-28)40-32-24-16-8-7-15-23(24)31(36-32)39-29-20-12-4-3-11-19(20)27(34-29)37-25;/h1-16H;/q-2;+2

InChI key

XCJYREBRNVKWGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jaron G Van Dijken et al.
Molecules (Basel, Switzerland), 17(9), 10119-10130 (2012-08-28)
We investigate the evolution of copper phthalocyanine thin films as they are etched with argon plasma. Significant morphological changes occur as a result of the ion bombardment; a planar surface quickly becomes an array of nanopillars which are less than
Richard Murdey et al.
The Journal of chemical physics, 134(23), 234702-234702 (2011-06-28)
The current flowing through a thin film of copper phthalocyanine vacuum deposited on a single crystal sapphire [0001] surface was measured during film growth from 0 to 93 nm. The results, expressed as conductance vs. nominal film thickness, indicate three
Kai Xiao et al.
Journal of the American Chemical Society, 135(9), 3680-3687 (2013-02-02)
The epitaxial growth and preferred molecular orientation of copper phthalocyanine (CuPc) molecules on graphene has been systematically investigated and compared with growth on Si substrates, demonstrating the role of surface-mediated interactions in determining molecular orientation. X-ray scattering and diffraction, scanning
Yu Li Huang et al.
Langmuir : the ACS journal of surfaces and colloids, 26(5), 3329-3334 (2010-02-24)
The self-assembly of the binary molecular system comprising copper(II) phthalocyanine (CuPc) and copper-hexadecafluoro-phthalocyanine (F(16)CuPc) on graphite has been investigated by in situ low-temperature scanning tunneling microscopy (LT-STM). The adsorption of this binary molecular system on graphite results in the formation
Jia Liu et al.
Journal of the American Chemical Society, 133(51), 21010-21015 (2011-11-24)
The bottom-up fabrication of surface hierarchical nanostructures is of great importance for the development of molecular nanostructures for chiral molecular recognition and enantioselective catalysis. Herein, we report the construction of a series of 2D chiral hierarchical structures by trinary molecular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service