Skip to Content
Merck
All Photos(2)

Key Documents

379549

Sigma-Aldrich

Iron(II) phthalocyanine

Dye content ~90 %

Synonym(s):

Phthalocyanine iron(II) salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H16FeN8
CAS Number:
Molecular Weight:
568.37
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

form

powder

composition

Dye content, ~90%

λmax

657 nm

SMILES string

[Fe]1n2c3nc4nc(nc5n1c(nc6nc(nc2c7ccccc37)c8ccccc68)c9ccccc59)c%10ccccc4%10

InChI

1S/C32H16N8.Fe/c1-2-10-18-17(9-1)25-33-26(18)38-28-21-13-5-6-14-22(21)30(35-28)40-32-24-16-8-7-15-23(24)31(36-32)39-29-20-12-4-3-11-19(20)27(34-29)37-25;/h1-16H;/q-2;+2

InChI key

MIINHRNQLVVCEW-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Fukui et al.
Journal of the American Geriatrics Society, 38(8), 889-892 (1990-08-01)
The authors designed a new pad for urinary incontinence. It is composed of a cloth of acrylonitrile copper sulfide and another cloth of iron-phthalocyanine. In vitro experiments showed that acrylonitrile copper sulfide cloth inhibited the growth of most bacteria causing
Anton A Ciucu et al.
Biosensors & bioelectronics, 18(2-3), 303-310 (2002-12-18)
A new highly sensitive amperometric method for the detection of organophosphorus compounds has been developed. The method is based on a ferophthalocyanine chemically modified carbon paste electrode coupled with acetylcholinesterase and choline oxidase co-immobilized onto the surface of a dialysis
H Ozaki et al.
Biochimica et biophysica acta, 955(2), 220-230 (1988-07-20)
Reactions of nitrosobenzene, phenyl isocyanide and their ring-substituted analogues with hemoglobin, ferrous phthalocyanine and a synthetic model compound of hemoglobin were investigated by optical, 1H-NMR and infrared spectroscopy. Complexes of chelated ferromesoheme, the model compound, with 2-methyl-, 2-ethyl, 2-isopropyl- or
R F Parton et al.
Nature, 370(6490), 541-544 (1994-08-18)
Many attempts have been made to mimic the catalytic oxidative properties of the enzyme cytochrome P-450. For homogeneous systems the mechanisms of oxidation can be readily determined but proper mimicry of the protein environment is difficult to achieve. Heterogeneous mimics

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service