Skip to Content
Merck
All Photos(1)

Documents

446653

Sigma-Aldrich

Copper(II) 1,2,3,4,8,9,10,11,15,16,17,18,22,23,24,25-hexadecafluoro-29H,31H-phthalocyanine

Dye content 80 %

Synonym(s):

F16CuPc

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32CuF16N8
CAS Number:
Molecular Weight:
863.92
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

powder

composition

Dye content, 80%

mp

>300 °C (lit.)

λmax

689 nm

SMILES string

Fc1c(F)c(F)c2c3nc(nc4n5[Cu]n6c(n3)c7c(F)c(F)c(F)c(F)c7c6nc8nc(nc5c9c(F)c(F)c(F)c(F)c49)c%10c(F)c(F)c(F)c(F)c8%10)c2c1F

InChI

1S/C32F16N8.Cu/c33-9-1-2(10(34)18(42)17(9)41)26-49-25(1)53-27-3-4(12(36)20(44)19(43)11(3)35)29(50-27)55-31-7-8(16(40)24(48)23(47)15(7)39)32(52-31)56-30-6-5(28(51-30)54-26)13(37)21(45)22(46)14(6)38;/q-2;+2

InChI key

FJAOBQORBYMRNO-UHFFFAOYSA-N

General description

Copper(II) 1,2,3,4,8,9,10,11,15,16,17,18,22,23,24,25-hexadecafluoro-29H,31H-phthalocyanine(F16CuPc) is an organic molecule that can be used as an electron transporting material with good air-stability and an electron mobility of 0.11 cm2/Vs . It is majorly used in a variety of energy conservation based applications.

Application

F16CuPc can be used as an electron transporting layer for semiconducting applications such as an electron accepting material for organic photovoltaic cells and as a catalyst in biomimetic sensors. It may also be used in the fabrication of organic thin film transistors(OTFTs) and quartz crystal microbalance(QCM) based sensors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetra-tert-butyl copper phthalocyanine-based QCM sensor for toluene detection in air at room temperature.
Kumar A, et al.
Sensors and Actuators B, Chemical, 210(1), 398-407 (2015)
Thickness-dependent structural transitions in fluorinated copper-phthalocyanine (F16CuPc) films.
de Oteyza DG, et al.
Journal of the American Chemical Society, 128(47), 15052-15053 (2006)
Copper hexadecafluorophthalocyanine (F16CuPc) as an electron accepting material in bilayer small molecule organic photovoltaic cells.
Yang JL, et al.
Organic Electronics, 11(8), 1399-1402 (2010)
Electroanalytical determination of bumetanide employing a biomimetic sensor for detection of doping in sports.
dos SR, et al.
Analytical Methods : Advancing Methods and Applications, 6(15), 5792-5798 (2014)
Organic photovoltaic cells using hexadecafluorophthalocyaninatocopper (F16CuPc) as electron acceptor material.
Jiang X, et al.
Chemical Physics Letters, 446(4-6), 329-332 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service