Skip to Content
Merck
All Photos(2)

Documents

546682

Sigma-Aldrich

Copper(II) phthalocyanine

Dye content >99 %

Synonym(s):

CuPc, Phthalocyanine blue, Pigment Blue 15

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H16CuN8
CAS Number:
Molecular Weight:
576.07
Colour Index Number:
74160
Beilstein:
4121848
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

composition

Dye content, >99%

λmax

678 nm

OLED Device Performance

ITO/CuPc/NPD/Alq3/C60/Mg:Ag

  • Color: green
  • Max. Luminance: 17200 Cd/m2

ITO/CuPc/NPD/Alq3/LiF/Al
  • Color: red
  • Max. Luminance: 2000 Cd/m2
  • Turn-On Voltage: 7.5 V

ITO/CuPc/NPD/CBP:FIrpic (6%)/BAlq3/LiF/Al
  • Color: blue
  • Max. EQE: 4.5 %

OPV Device Performance

ITO/CuPc/PTCDA/In

  • Short-circuit current density (Jsc): 53.1 mA/cm2
  • Open-circuit voltage (Voc): 0.57 V
  • Fill Factor (FF): 0.61
  • Power Conversion Efficiency (PCE): 4.2 %

ITO/PEDOT:PSS/CuPc/C60/BCP/Al
  • Short-circuit current density (Jsc): 18.8 mA/cm2
  • Open-circuit voltage (Voc): 0.58 V
  • Fill Factor (FF): 0.52
  • Power Conversion Efficiency (PCE): 3.6 %

SMILES string

c1ccc2c(c1)C3=NC4=[N@@H]5C(=Nc6n7c(N=C8c9ccccc9C%10=[N@@H]8[Cu]57N3C2=N%10)c%11ccccc6%11)c%12ccccc4%12

InChI

1S/C32H16N8.Cu/c1-2-10-18-17(9-1)25-33-26(18)38-28-21-13-5-6-14-22(21)30(35-28)40-32-24-16-8-7-15-23(24)31(36-32)39-29-20-12-4-3-11-19(20)27(34-29)37-25;/h1-16H;/q-2;+2

InChI key

XCJYREBRNVKWGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • A facile molecularly engineered copper (II) phthalocyanine as hole transport material for planar perovskite solar cells with enhanced performance and stability: This study introduces a modified copper(II) phthalocyanine that enhances the performance and stability of perovskite solar cells (Yang et al., 2017).

  • Dopant-free methoxy substituted copper (II) phthalocyanine for highly efficient and stable perovskite solar cells: Discusses the synthesis and application of a methoxy-substituted copper(II) phthalocyanine, improving the efficiency and stability of perovskite solar cells (Ding et al., 2020).

  • Highly efficient dye-sensitized solar cells based on metal-free and copper (II) phthalocyanine bearing 2-phenylphenoxy moiety: Examines novel phthalocyanines for use in dye-sensitized solar cells, focusing on their synthesis and photophysical properties (Ali et al., 2016).

  • Operando HERFD-XANES and surface sensitive Delta mu analyses identify the structural evolution of copper (II) phthalocyanine for electroreduction of CO2: This research uses advanced spectroscopic techniques to explore the structural changes in copper(II) phthalocyanine during CO2 electroreduction (Mei et al., 2022).

  • New dye sensitized photocatalysts: Copper (II)-phthalocyanine/TiO2 nanocomposite for water remediation: Studies a copper(II) phthalocyanine-TiO2 composite as a photocatalyst for water remediation, showing its effectiveness in degrading pollutants under light irradiation (Albay et al., 2016).

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kai Xiao et al.
Journal of the American Chemical Society, 135(9), 3680-3687 (2013-02-02)
The epitaxial growth and preferred molecular orientation of copper phthalocyanine (CuPc) molecules on graphene has been systematically investigated and compared with growth on Si substrates, demonstrating the role of surface-mediated interactions in determining molecular orientation. X-ray scattering and diffraction, scanning
Jaron G Van Dijken et al.
Molecules (Basel, Switzerland), 17(9), 10119-10130 (2012-08-28)
We investigate the evolution of copper phthalocyanine thin films as they are etched with argon plasma. Significant morphological changes occur as a result of the ion bombardment; a planar surface quickly becomes an array of nanopillars which are less than
Richard Murdey et al.
The Journal of chemical physics, 134(23), 234702-234702 (2011-06-28)
The current flowing through a thin film of copper phthalocyanine vacuum deposited on a single crystal sapphire [0001] surface was measured during film growth from 0 to 93 nm. The results, expressed as conductance vs. nominal film thickness, indicate three
Medhat A Shaibat et al.
The journal of physical chemistry. B, 114(13), 4400-4406 (2010-03-17)
Cu(II)(phthalocyanine) (CuPc) is broadly utilized as an archetypal molecular semiconductor and is the most widely used blue printing pigment. CuPc crystallizes in six different forms; the chemical and physical properties are substantially modulated by its molecular packing among these polymorphs.
Jiannan Dong et al.
Langmuir : the ACS journal of surfaces and colloids, 26(10), 6995-7006 (2010-01-16)
The dispersion stability and the zeta potentials of nonspherical crystalline (beta-form) copper phthalocyanine (CuPc) particles of hydrodynamic diameter d(h) approximately = 90 nm were investigated at 25 degrees C in water and in aqueous solutions of NaNO(3). The electrolyte concentrations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service