Merck
  • In situ conductance measurements of copper phthalocyanine thin film growth on sapphire [0001].
In situ conductance measurements of copper phthalocyanine thin film growth on sapphire [0001].
The Journal of chemical physics (2011-06-28)
Richard Murdey, Naoki Sato
ABSTRACT
The current flowing through a thin film of copper phthalocyanine vacuum deposited on a single crystal sapphire [0001] surface was measured during film growth from 0 to 93 nm. The results, expressed as conductance vs. nominal film thickness, indicate three distinct film growth regions. Conductive material forms below about 5 nm and again above 35 nm, but in the intermediate thicknesses the film conductance was observed to decrease with increasing film thickness. With the aid of ac-AFM topology images taken ex situ, the conductance results are explained based on the Stranski-Krastanov (2D + 3D) film growth mechanism, in which the formation of a thin wetting layer is followed by the growth of discrete islands that eventually coalesce into an interpenetrating, conductive network.
Sigma-Aldrich
Copper(II) phthalocyanine, Dye content >99 %
Sigma-Aldrich
Copper(II) phthalocyanine, sublimed grade, Dye content 99 %
Sigma-Aldrich
Copper(II) phthalocyanine, β-form, Dye content 90 %
Sigma-Aldrich
Copper(II) phthalocyanine, triple-sublimed grade, >99.95% trace metals basis