Skip to Content
Merck
All Photos(2)

Key Documents

637149

Sigma-Aldrich

Iron nickel oxide

nanopowder, <50 nm particle size (APS), ≥98% trace metals basis

Synonym(s):

Diiron nickel tetraoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe2NiO4
CAS Number:
Molecular Weight:
234.38
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

≥98% trace metals basis

form

nanopowder

particle size

<50 nm (APS)

density

5.368 g/mL at 25 °C (lit.)

bulk density

0.89 g/mL

application(s)

battery manufacturing

SMILES string

O=[Ni].O=[Fe]O[Fe]=O

InChI

1S/2Fe.Ni.4O

InChI key

NQNBVCBUOCNRFZ-UHFFFAOYSA-N

General description

Iron nickel oxide (Fe2O3/NiO) nanopowder is a class of nanomaterial with properties such as superior magnetic, electrical and selective absorption. It can be used as an additive in the formation of membranes for water treatment.

Application

Fe2O3/NiO can be used for applications such as:
  • fabrication of magnetic nanofiber for data storage and transfer
  • electrocatalysts for the splitting of alkaline water
  • formation of composite nanofibers for efficient water oxidation electrocatalysis

Physical form

Nearly spherical crystalline morphology

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrospinning synthesis of bimetallic nickel-iron oxide/carbon composite nanofibers for efficient water oxidation electrocatalysis
Chen H, et al.
ChemCatChem, 8(5), 992-1000 (2016)
Iron-doped nickel oxide nanocrystals as highly efficient electrocatalysts for alkaline water splitting
Fominykh K, et al.
ACS Nano, 9(5), 5180-5188 (2015)
Xing Guo et al.
Sensors (Basel, Switzerland), 20(18) (2020-09-20)
For the first time, a novel NiFe2O4/paper-based magnetoelastic (ME) biosensor was developed for rapid, sensitive, and portable detection of human serum albumin (HSA). Due to the uniquely magnetoelastic effect of NiFe2O4 nanoparticles and the excellent mechanical properties of the paper
Magnetic nanofiber mats for data storage and transfer
Dopke C, et al.
Nanomaterials, 9(1), 92-92 (2019)
Preparation and characterization of ion-selective polyvinyl chloride based heterogeneous cation exchange membrane modified by magnetic iron-nickel oxide nanoparticles
Hosseini SM, et al.
Desalination, 284, 191-199 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service