Skip to Content
Merck
All Photos(3)

Key Documents

641669

Sigma-Aldrich

Nickel zinc iron oxide

nanopowder, <100 nm particle size (BET), ≥99% trace metals basis

Synonym(s):

Nickel zinc ferrite, Zinc nickel ferrite, Zinc nickel iron oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NiZnFe4O4
CAS Number:
Molecular Weight:
411.46
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99% trace metals basis

form

nanopowder

particle size

<100 nm (BET)

density

2.81 g/mL at 25 °C (lit.)

SMILES string

[Ni++].[Cu++].[O-][Fe]=O.[O-][Fe]=O.[O-][Fe]=O.[O-][Fe]=O

InChI

1S/2Fe.Ni.4O.Zn

InChI key

XGKOQBJLEIRRCP-UHFFFAOYSA-N

Legal Information

Product of Engi-Mat Co.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

P Majewski et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 14(26), 7961-7968 (2008-07-12)
We report on the spontaneous covalent growth of monomolecular adlayers on mixed nickel-zinc nanoferrite colloidal suspensions (ferrofluids). Synthesized nanoparticles were subjected to surface modification by means of acid chloride chemistry, leading to the formation of covalent bonds between the hydroxy
[Embryotoxic action of nickel-zinc ferrite in body uptake by inhalation].
N N Govorunova et al.
Gigiena i sanitariia, (5)(5), 79-80 (1984-05-01)
[Changes in the blood protein spectrum of white rats exposed to inhalational uptake of ferrites].
V D Aleshina et al.
Gigiena i sanitariia, (6)(6), 63-65 (1986-06-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service