Skip to Content
Merck
All Photos(2)

Key Documents

773352

Sigma-Aldrich

Cobalt iron oxide

nanopowder, 30 nm particle size (TEM), 99% trace metals basis

Synonym(s):

Cobalt diiron tetraoxide, Cobalt ferrate, Cobalt ferrite, Cobalt iron spinel

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CoFe2O4
CAS Number:
Molecular Weight:
234.62
EC Number:
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23
form:
nanopowder

Quality Level

Assay

99% trace metals basis

form

nanopowder

particle size

30 nm (TEM)

application(s)

battery manufacturing

SMILES string

[Co+2].[Co+2].[Fe+3].[Fe+3].[O-2].[O-2].[O-2].[O-2].[O-2]

InChI

1S/2Co.2Fe.5O/q2*+2;2*+3;5*-2

InChI key

XNGOCNMGDFPQTP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cobalt Iron Oxide is an important nanoscale magnetic material with applications in molecular imaging† and drug delivery

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Friedrich Waag et al.
Scientific reports, 7(1), 13161-13161 (2017-10-17)
Highly active, structurally disordered CoFe
Multifunctional nanoparticles possessing a "magnetic motor effect" for drug or gene delivery.
Tae-Jong Yoon et al.
Angewandte Chemie (International ed. in English), 44(7), 1068-1071 (2005-01-07)
Jae-Hyun Lee et al.
Nature medicine, 13(1), 95-99 (2006-12-26)
Successful development of ultra-sensitive molecular imaging nanoprobes for the detection of targeted biological objects is a challenging task. Although magnetic nanoprobes have the potential to perform such a role, the results from probes that are currently available have been far
Maqusood Ahamed et al.
Colloids and surfaces. B, Biointerfaces, 148, 665-673 (2016-11-05)
Cobalt iron oxide (CoFe

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service