Skip to Content
Merck
All Photos(2)

Documents

544884

Sigma-Aldrich

Iron(III) oxide

nanopowder, <50 nm particle size (BET)

Synonym(s):

Ferric oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Fe2O3
CAS Number:
Molecular Weight:
159.69
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

description

crystalline (primarily γ)

form

nanopowder

surface area

50-245 m2/g

particle size

<50 nm (BET)

application(s)

battery manufacturing

SMILES string

O=[Fe]O[Fe]=O

InChI

1S/2Fe.3O

InChI key

JEIPFZHSYJVQDO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Iron(III) oxide nanopowder is a fine powder with a particle size of less than 50 nm. It is a red or black solid compound made up of iron and oxygen. It is also known as hematite or ferric oxide. It is a naturally occurring mineral that can also be synthesized in the laboratory. Iron(III) oxide has a number of useful physical properties. It has a high refractive index and is opaque, making it useful as a pigment in paints in inks. Iron(III) oxide is also catalytically active and weakly ferromagnetic at room temperature.

Application

Iron(III) oxide nanopowder has a number of uses due to its magnetic and catalytic properties. It is used in the production of magnetic recording media such as magnetic tapes and disks. It is also used as a catalyst in the production of chemicals, including the production of gasoline and plastics and in environmental remediation.

Features and Benefits

  • High theoretical specific capacity
  • Biocompatibility
  • Ease of coating and modification
  • Non-toxicity

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junho Han et al.
Scientific reports, 9(1), 6130-6130 (2019-04-18)
Recent developments in analytics using infrared spectroscopy have enabled us to identify the adsorption mechanism at interfaces, but such methods are applicable only for simple systems. In this study, the preferential adsorption of phosphate on binary goethite and maghaemite was
Daniel Matatagui et al.
Sensors (Basel, Switzerland), 19(24) (2019-12-11)
A portable electronic nose based on surface acoustic wave (SAW) sensors is proposed in this work to detect toxic chemicals, which have a great potential to threaten the surrounding natural environment or adversely affect the health of people. We want
Hokuto Fuse et al.
Nanomaterials (Basel, Switzerland), 9(2) (2019-02-06)
Submicrometre spherical particles made of Au and Fe can be fabricated by pulsed-laser melting in liquid (PLML) using a mixture of Au and iron oxide nanoparticles as the raw particles dispersed in ethanol, although the detailed formation mechanism has not
Yangyang Yang et al.
Ecotoxicology and environmental safety, 148, 89-96 (2017-10-17)
The behaviors of nanoparticles rely on the aqueous condition such as natural organic matter (NOM). Therefore the presence of NOM would influence the interaction of nanoparticles with other substances possibly. Here, microcystin-LR (MC-LR) adsorption on iron oxide nanoparticles (IONPs) was
Alice Panariti et al.
Journal of biomedical nanotechnology, 9(9), 1556-1569 (2013-08-29)
Magnetic nanoparticles have emerged as important players in current research in modern medicine since they can be used in medicine for diagnosis and/or therapeutic treatment of diseases. Among many therapeutic applications of iron-based nanoparticles, drug delivery and photothermal therapy are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service