Skip to Content
Merck
All Photos(3)

Key Documents

697230

Sigma-Aldrich

[1,1′-Bis(diphenylphosphino)ferrocene]dichloropalladium(II)

[1,1′-Bis(diphenylphosphino)ferrocene]dichloropalladium(II)

Synonym(s):

Pd(dppf)Cl2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C17H14P)2Fe · PdCl2
CAS Number:
Molecular Weight:
731.70
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

form

powder

Quality Level

reaction suitability

core: palladium
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Buchwald-Hartwig Cross Coupling Reaction

mp

266-283 °C (lit.)

SMILES string

[Fe].Cl[Pd]Cl.[CH]1[CH][CH][C]([CH]1)P(c2ccccc2)c3ccccc3.[CH]4[CH][CH][C]([CH]4)P(c5ccccc5)c6ccccc6

InChI

1S/2C17H14P.2ClH.Fe.Pd/c2*1-3-9-15(10-4-1)18(17-13-7-8-14-17)16-11-5-2-6-12-16;;;;/h2*1-14H;2*1H;;/q;;;;;+2/p-2

InChI key

JCWIWBWXCVGEAN-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for C-C and C-N coupling reaction.
Pd(dppf)Cl2 may be used as an effective palladium catalyst in the following reactions:
  • Cross-coupling of sec-alkyl and n-alkyl Grignard reagents with high yield and selectivity.[1]
  • Suzuki coupling of aryl boronic esters with [11C]methyl iodide to form functionalized [11C]toluene derivatives.[2]
  • Kumada cross-coupling of 1,3,5-tribromobenzene with Grignard reagents to form star-shaped oligothiophenes.[3]
For small scale and high throughput uses, product is also available as ChemBeads (919780)

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dichloro [1,1?-bis (diphenylphosphino) ferrocene] palladium (II): an effective catalyst for cross-coupling reaction of a secondary alkyl grignard reagent with organic halides.
Hayashi T
Tetrahedron Letters, 20(21), 1871-1874 (1979)
Shirbin, S. J. et al.
Tetrahedron Letters, 51, 2971-2971 (2010)
Star?Shaped Oligothiophenes for Solution?Processible Organic Field?Effect Transistors.
Ponomarenko S
Advances in Functional Materials, 13(8), 591-596 (2003)
Paul A Allegretti et al.
Bioorganic & medicinal chemistry, 28(1), 115193-115193 (2019-11-24)
Small molecule stimulation of β-cell regeneration has emerged as a promising therapeutic strategy for diabetes. Although chemical inhibition of dual specificity tyrosine-phosphorylation-regulated kinase 1A (DYRK1A) is sufficient to enhance β-cell replication, current lead compounds have inadequate cellular potency for in
An improved synthesis of substituted [11C] toluenes via Suzuki coupling with [11C] methyl iodide
Hostetler E
Journal of Labelled Compounds & Radiopharmaceuticals, 48(9), 629-634 (2005)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

Active Filters

  1. 2 Ratings-Only Reviews

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service