Skip to Content
Merck
All Photos(3)

Key Documents

60811

Sigma-Aldrich

Cobalt carbonyl

moistened with hexane (hexane 1-10%), ≥90% (Co)

Synonym(s):

Cobalt tetracarbonyl dimer, Di-μ-carbonylhexacarbonyldicobal, Octacarbonyldicobalt, Dicobalt octacarbonyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Co2(CO)8
CAS Number:
Molecular Weight:
341.95
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥90% (Co)

form

solid

quality

moistened with hexane (hexane 1-10%)

reaction suitability

core: cobalt
reagent type: catalyst

storage temp.

2-8°C

SMILES string

[Co++].[Co++].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]

InChI

1S/8CO.2Co/c8*1-2;;/q;;;;;;;;2*+2

InChI key

MQIKJSYMMJWAMP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Cobalt carbonyl [Co2(CO)8] is commonly used as a catalyst in the hydroformylation (oxo reaction) of alkenes.
  • Along with pyridine, it can be used as a catalyst in the carboxylation of alkenes into corresponding acids and esters.
  • It is employed as a key precursor in the preparation of cobalt platinum (CoPt3), cobalt sulfide (Co3S4) and cobalt selenide (CoSe2) nanocrystals.
  • It is also used as a reagent in Pauson-Khand cyclizations and Nicholas reaction.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Repr. 2 - Self-heat. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Inhalation

Target Organs

Nervous system

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

-9.4 °F

Flash Point(C)

-23 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dicobalt octacarbonyl.
Fischer S.
Synlett, 2002(09), 1558-1559 (2002)
In situ free-surfactant synthesis and ORR-electrochemistry of carbon-supported Co3S4 and CoSe2 nanoparticles.
Feng Y, et al.
Chemistry of Materials, 20(1), 26-28 (2007)
A Lewis acid-mediated version of the Nicholas reaction: Synthesis of syn-alkylated products and cobalt-complexed cycloalkynes.
Schreiber SL, et al.
Journal of the American Chemical Society, 108(11), 3128-3130 (1986)
M Varón et al.
Scientific reports, 3, 1234-1234 (2013-02-08)
Magnetostatic (dipolar) interactions between nanoparticles promise to open new ways to design nanocrystalline magnetic materials and devices if the collective magnetic properties can be controlled at the nanoparticle level. Magnetic dipolar interactions are sufficiently strong to sustain magnetic order at
Octacarbonyldicobalt.
Encyclopedia of Reagents for Organic Synthesis, Second Edition, 1-26 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service