Skip to Content
Merck
All Photos(3)

Key Documents

225487

Sigma-Aldrich

Cyclopentadienyl iron(II) dicarbonyl dimer

greener alternative

99%

Synonym(s):

Dicarbonylcyclopentadienyliron dimer, Dicyclopentadienyliron dicarbonyl dimer, Tetracarbonylbis(cyclopentadienyl)diiron, Bis(cyclopentadienyl)tetracarbonyl-diiron, Di(cyclopentadienyl)tetracarbonyl-diiron

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10Fe2O4
CAS Number:
Molecular Weight:
353.92
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

reaction suitability

core: iron
reagent type: catalyst

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

194 °C (dec.) (lit.)

greener alternative category

SMILES string

[Fe][Fe].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[CH]1[CH][CH][CH][CH]1.[CH]2[CH][CH][CH][CH]2

InChI

1S/2C5H5.4CO.2Fe/c2*1-2-4-5-3-1;4*1-2;;/h2*1-5H;;;;;;

InChI key

DHLWUDJVLCWRGH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

Dinuclear metal carbonyl catalyst for a variety of carbon-carbon bond forming reactions; radical initiator; precursor to alkyland acyliron complexes useful in organic synthesis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Davis, R.; Khazal, N. M. S. et al.
Journal of Organometallic Chemistry, 397, 51-51 (1990)
Brookhart, M.; Liu, Y. et al.
Journal of the American Chemical Society, 113, 927-927 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service