Skip to Content
Merck
All Photos(4)

Key Documents

245011

Sigma-Aldrich

Triruthenium dodecacarbonyl

99%

Synonym(s):

Ruthenium carbonyl, tri-Ruthenium dodecacarbonyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ru3(CO)12
CAS Number:
Molecular Weight:
639.33
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

crystals

reaction suitability

core: ruthenium
reaction type: C-H Activation
reagent type: catalyst

SMILES string

[Ru].[Ru].[Ru].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]

InChI

1S/12CO.3Ru/c12*1-2;;;

InChI key

NQZFAUXPNWSLBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Atomic number of base material: 44 Ruthenium

Application

Carbonyl cluster precursor and H-transfer catalyst. Used in the reductive carbonylation of aromatic nitro compounds to carbamates. The phosphine-stabilized carbonyl cluster has been tethered to oxide supports. Applied in improved catalysis of the allylic amination of unactivated olefins by nitroarenes.
Catalyst used in a [3+2+1] carbonylative cycloaddition of silylacteylenes and α,ß-unsaturated ketones providing good yields of α-pyrones.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Inhalation

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takahide Fukuyama et al.
Organic letters, 9(4), 587-589 (2007-01-25)
Ruthenium catalyzes a carbonylative [3+2+1] cycloaddition, using silylacetylenes, alpha,beta-unsaturated ketones, and CO as the starting materials, providing the new method for the synthesis of tetrasubstituted alpha-pyrones. In this reaction, the carbonyl group and alpha-carbon of vinyl ketones are incorporated as
Ragaini, F. et al.
Organometallics, 18, 928-928 (1999)
J. Chem. Soc., Dalton Trans., 2649-2649 (1988)
Ragaini, F. et al.
Organometallics, 18, 929-929 (1999)
Journal of the American Chemical Society, 106, 4647-4647 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service