Skip to Content
Merck
All Photos(2)

Key Documents

247650

Sigma-Aldrich

Hexarhodium(0) hexadecacarbonyl

Rh 57-60 % (approx.)

Synonym(s):

Rhodium(0) carbonyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Rh6(CO)16
CAS Number:
Molecular Weight:
1065.59
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

composition

Rh, 57-60% (approx.)

reaction suitability

core: rhodium
reagent type: catalyst

SMILES string

[Rh].[Rh].[Rh].[Rh].[Rh].[Rh].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]

InChI

1S/16CO.6Rh/c16*1-2;;;;;;

InChI key

SZQABOJVTZVBHE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for hydrogenation of alkenes, hydroformylations ofalkenes; reducing agent for aldehydes, ketones, and nitro groups; catalyst for carbenoid reactions of diazo compounds
Widely used organometallic catalyst. Applications include the hydroformylation of olefins.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Crotti, C.; Cenini, S.; Rindone, B. et al.
Journal of the Chemical Society. Chemical Communications, 784-784 (1986)
Journal of Materials Chemistry, 5, 75-75 (1995)
Murahashi, S.; Imada, Y.; Hirai, Y.
Bulletin of the Chemical Society of Japan, 62, 2968-2968 (1989)
Journal of the American Ceramic Society. American Ceramic Society, 78, 1179-1179 (1995)
Ojima, I.; Kato, K.; Okabe, M.; Fuchikami, T.
Journal of the American Chemical Society, 109, 7714-7714 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service