Skip to Content
Merck
All Photos(1)

Key Documents

49159

Millipore

D-(+)-Glucose monohydrate

D-(+)-Glucose monohydrate

suitable for microbiology, ≥99.0%

Synonym(s):

Dextrose monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6 · H2O
CAS Number:
Molecular Weight:
198.17
Beilstein:
5250278
EC Number:
MDL number:
UNSPSC Code:
41106212
PubChem Substance ID:
NACRES:
NA.85
Pricing and availability is not currently available.

Quality Level

Assay

≥99.0% (sum of enantiomers, HPLC)
≥99.0%

form

powder

optical activity

[α]20/D +49±3°, 3 hr, c = 10% in H2O

shelf life

limited shelf life, expiry date on the label

packaging

pkg of 1 kg
pkg of 5 kg

storage condition

(Tightly closed)

ign. residue

≤0.1% (as SO4)

pH

6-7 (20 °C, 100 g/L in water)

solubility

H2O: 0.1 g/mL, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

As: ≤0.1 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

application(s)

microbiology

SMILES string

O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C=O

InChI

1S/C6H12O6.H2O/c7-1-3(9)5(11)6(12)4(10)2-8;/h1,3-6,8-12H,2H2;1H2/t3-,4+,5+,6+;/m0./s1

InChI key

SPFMQWBKVUQXJV-BTVCFUMJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

D-(+)-Glucose monohydrate, commonly known as glucose or dextrose, has several microbiological applications. It is used as a carbon source in microbial culture media to promote the growth and propagation of variety of microorganisms. Glucose is a simple sugar and a primary energy source for bacterial cell metabolism. Glucose monohydrate can also be used as a supplement in media to enhance the growth and productivity of microorganisms used in fermentation processes, for example, in the production of antibiotics, organic acids, and enzymes. It plays an important role in diauxic growth, which refers to the phenomenon in which bacterial cells preferentially utilize glucose as a carbon source before switching to other energy sources. In addition, glucose is a common component in numerous selective and differential media formulations used in the identification and characterization of microorganisms.

Application

D-(+)-Glucose monohydrate is commonly used as an ingredient in many microbial culture media for the growth and identification of bacteria, yeasts, and molds from various pharmaceutical, environmental, and food and beverage samples.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cicera R Lazzarotto et al.
Nature protocols, 13(11), 2615-2642 (2018-10-21)
Circularization for in vitro reporting of cleavage effects by sequencing (CIRCLE-seq) is a sensitive and unbiased method for defining the genome-wide activity (on-target and off-target) of CRISPR-Cas9 nucleases by selective sequencing of nuclease-cleaved genomic DNA (gDNA). Here, we describe a
Daphne H E W Huberts et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(32), 11727-11731 (2014-07-30)
Calorie restriction (CR) is often described as the most robust manner to extend lifespan in a large variety of organisms. Hence, considerable research effort is directed toward understanding the mechanisms underlying CR, especially in the yeast Saccharomyces cerevisiae. However, the
Paola Rusmini et al.
Autophagy, 15(4), 631-651 (2018-10-20)
Macroautophagy/autophagy, a defense mechanism against aberrant stresses, in neurons counteracts aggregate-prone misfolded protein toxicity. Autophagy induction might be beneficial in neurodegenerative diseases (NDs). The natural compound trehalose promotes autophagy via TFEB (transcription factor EB), ameliorating disease phenotype in multiple ND
Antonio Marsella et al.
Cell reports, 34(13), 108906-108906 (2021-04-01)
The Mre11-Rad50-Xrs2 (MRX) complex detects and processes DNA double-strand breaks (DSBs). Its DNA binding and processing activities are regulated by transitions between an ATP-bound state and a post-hydrolysis cutting state that is nucleolytically active. Mre11 endonuclease activity is stimulated by
Maiken Mellergaard et al.
The Journal of biological chemistry, 295(33), 11803-11821 (2020-07-02)
Staphylococcus aureus is among the leading causes of bacterial infections worldwide. The pathogenicity and establishment of S. aureus infections are tightly linked to its ability to modulate host immunity. Persistent infections are often associated with mutant staphylococcal strains that have

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service