Skip to Content
Merck
All Photos(1)

Documents

514012

Sigma-Aldrich

1-Azido-1-deoxy-β-D-lactopyranoside

97%

Synonym(s):

1-Azido-1-deoxy-beta-D-lactopyranoside, 1-Azido-1-deoxy-beta-D-lactose, 4-O-beta-D-Galactopyranosyl-beta-D-glucopyranosyl azide, beta-Lactosyl azide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H21N3O10
CAS Number:
Molecular Weight:
367.31
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

optical activity

[α]20/D +14.8°, c = 1% in DMSO

reaction suitability

reaction type: click chemistry

mp

120 °C (lit.)

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O[C@@H]2CO)N=[N+]=[N-])[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C12H21N3O10/c13-15-14-11-8(21)7(20)10(4(2-17)23-11)25-12-9(22)6(19)5(18)3(1-16)24-12/h3-12,16-22H,1-2H2/t3-,4-,5+,6+,7-,8-,9-,10-,11-,12+/m1/s1

InChI key

FJOYNMRZUBUGGP-DCSYEGIMSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service