Skip to Content
Merck
All Photos(3)

Key Documents

511447

Sigma-Aldrich

Phenylbis(2,4,6-trimethylbenzoyl)phosphine oxide

97%, powder

Synonym(s):

BAPOs, Bisacylphosphine oxides

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3C6H2CO]2P(O)C6H5
CAS Number:
Molecular Weight:
418.46
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

powder
powder, crystals or chunks

mp

131-135 °C (lit.)

solubility

acetone, acetonitrile, toluene, and hexanediol diacrylate: soluble

SMILES string

Cc1cc(C)c(c(C)c1)C(=O)P(=O)(C(=O)c2c(C)cc(C)cc2C)c3ccccc3

InChI

1S/C26H27O3P/c1-16-12-18(3)23(19(4)13-16)25(27)30(29,22-10-8-7-9-11-22)26(28)24-20(5)14-17(2)15-21(24)6/h7-15H,1-6H3

InChI key

GUCYFKSBFREPBC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phenylbis(2,4,6-trimethylbenzoyl) phosphineoxide (BAPO) is potential photoinitiator widely used in the preparation ofdental composites.

Application

Phenylbis(2,4,6-trimethylbenzoyl) phosphine oxide can be used as a photoinitiator for:
  • Radical polymerization of dental resins. BAPO enhances the polymerization rate and conversion compared to other initiators.
  • Preparation of divinylbenzene modified polymer-based ceramic materials for high temperature sensor applications.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Federico Lancia et al.
Nature communications, 10(1), 4819-4819 (2019-10-28)
The motion of artificial molecular machines has been amplified into the shape transformation of polymer materials that have been compared to muscles, where mechanically active molecules work together to produce a contraction. In spite of this progress, harnessing cooperative molecular
Maria Salem Ibrahim et al.
Materials (Basel, Switzerland), 11(9) (2018-08-29)
Dental sealants with antibacterial and remineralizing properties are promising for caries prevention among children and adolescents. The application of nanotechnology and polymer development have enabled nanoparticles of amorphous calcium phosphate (NACP) and dimethylaminohexadecyl methacrylate (DMAHDM) to emerge as anti-caries strategies
Mauricio Castellanos et al.
The journal of adhesive dentistry, 21(1), 67-76 (2019-02-26)
To evaluate the light transmittance of ceramic veneers of different thicknesses and verify their influence on the degree of conversion, color stability, and dentin bond strength of light-curing resin cements containing different photoinitiator systems. Experimental resin cements were fabricated containing
Samantha Molina-Gutiérrez et al.
Molecules (Basel, Switzerland), 25(15) (2020-08-06)
Biobased monomers have been used to replace their petroleum counterparts in the synthesis of polymers that are aimed at different applications. However, environmentally friendly polymerization processes are also essential to guarantee greener materials. Thus, photoinduced polymerization, which is low-energy consuming
Michael J Beauchamp et al.
Micromachines, 9(7) (2018-11-15)
Interest has grown in recent years to leverage the possibilities offered by three-dimensional (3D) printing, such as rapid iterative changes; the ability to more fully use 3D device volume; and ease of fabrication, especially as it relates to the creation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service