Skip to Content
Merck
All Photos(3)

Key Documents

381489

Sigma-Aldrich

Trimethylolpropane tris(3-mercaptopropionate)

≥95.0%

Synonym(s):

1,1,1-Trimethylolpropane-tris(3-mercaptopropionate), 2,2-Bis(3-sulfanylpropanoyloxymethyl)butyl 3-sulfanylpropanoate, Trimethylolpropane tris(β-mercaptopropionate), Trimethylolpropane tris(3-mercaptopropanoate), Trimethylolpropane tris(3-mercaptopropropionate)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HSCH2CH2CO2CH2)3CC2H5
CAS Number:
Molecular Weight:
398.56
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0%

refractive index

n20/D 1.518 (lit.)

bp

220 °C/0.3 mmHg (lit.)

density

1.21 g/mL at 25 °C (lit.)

functional group

ester
thiol

SMILES string

CCC(COC(=O)CCS)(COC(=O)CCS)COC(=O)CCS

InChI

1S/C15H26O6S3/c1-2-15(9-19-12(16)3-6-22,10-20-13(17)4-7-23)11-21-14(18)5-8-24/h22-24H,2-11H2,1H3

InChI key

IMQFZQVZKBIPCQ-UHFFFAOYSA-N

General description

Trimethylolpropane tris(3-mercaptopropionate) is reported as a multi-thiol crosslinking reagent.

Application

Trimethylolpropane tris(3-mercaptopropionate) may be used in the following studies:
  • Preparation of hydrogels, via crosslinking inulin derivatives.
  • Construction of porous hybrid monolithic materials.
  • As monomer for the preparation of a new thiol-ene based polymeric fluorescence sensor, via photo initiated polymerization.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giovanna Pitarresi et al.
Macromolecular bioscience, 8(10), 891-902 (2008-06-26)
Inulin was chosen as a starting polymer for biocompatible, pH-sensitive and biodegradable hydrogels. Three INUDVSA-TT hydrogels were obtained by crosslinking inulin derivatives with trimethylolpropane tris(3-mercaptopropionate) under varying conditions. The resulting hydrogels were cell compatible, as demonstrated by MTS and trypan
Soner Cubuk et al.
Analytica chimica acta, 812, 215-221 (2014-02-05)
This study describes the preparation and the characterization of a new thiol-ene based polymeric fluorescence sensor by photo initiated polymerization of trimethylolpropane tris(3-mercaptopropionate), 2-hydroxyethylacrylate, and 2,4,6-triallyloxy-1,3,5-triazine which are used as monomers and also a photo initiator (2,2-dimethoxy-2-phenylacetophenone) for its usage
Sarah A Richardson et al.
Macromolecular bioscience, 19(2), e1800351-e1800351 (2018-12-15)
A novel strategy for the surface functionalization of emulsion-templated highly porous (polyHIPE) materials as well as its application to in vitro 3D cell culture is presented. A heterobifunctional linker that consists of an amine-reactive N-hydroxysuccinimide ester and a photoactivatable nitrophenyl
Dailyn Guzmán et al.
Polymers, 12(2) (2020-02-09)
The pure trifunctional glycidyl monomer from phloroglucinol (3EPO-Ph) was synthesized and used as feedstock in the preparation of novel bio-based thermosets by thiol-epoxy curing. The monomer was crosslinked with different commercially available thiols: tetrafunctional thiol (PETMP), trifunctional thiol (TTMP) and
Alexandra N Tsoras et al.
Frontiers in immunology, 11, 1547-1547 (2020-08-28)
Peptide subunit vaccines increase safety by reducing the risk of off-target responses and improving the specificity of the induced adaptive immune response. The immunogenicity of most soluble peptides, however, is often insufficient to produce robust and lasting immunity. Many biomaterials

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service