Skip to Content
Merck
All Photos(3)

Key Documents

405612

Sigma-Aldrich

1-Hydroxycyclohexyl phenyl ketone

99%

Synonym(s):

(1-Hydroxycyclohexyl)phenylmethanone, 1-Benzoyl-1-hydroxycyclohexane, 1-Benzoylcyclohexanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H10COC6H5
CAS Number:
Molecular Weight:
204.26
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99%

bp

175 °C/15 mmHg (lit.)

mp

47-50 °C (lit.)

SMILES string

OC1(CCCCC1)C(=O)c2ccccc2

InChI

1S/C13H16O2/c14-12(11-7-3-1-4-8-11)13(15)9-5-2-6-10-13/h1,3-4,7-8,15H,2,5-6,9-10H2

InChI key

QNODIIQQMGDSEF-UHFFFAOYSA-N

General description

1-Hydroxycyclohexyl phenyl ketone is a photoinitiator (PI) molecule that can be used in chain transfer polymerization. PI can be incorporated in the polymeric matrix by the addition of a chromophore as a pendant group. It may be used as a component that facilitates UV curing and also as a base material in the formation of the block and grafted copolymers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

327.2 °F - closed cup

Flash Point(C)

164 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of novel well-defined end-functional macrophotoinitiator of poly (MMA) by ATRP
Degirmenci M
Journal of Macromolecular Science, Part A: Pure and Applied Chemistry, 42(1), 21-30 (2005)
Jian Teng et al.
Carbohydrate polymers, 179, 244-251 (2017-11-08)
The ultra-high mechanical, biocompatible and biodegradable porous regenerated cellulose/poly(ethylene glycol) (RC/PEG) composites with double network structure were fabricated via an simple method to dissolve cellulose followed by UV irradiation. The porous structure of RC/PEG was sensitively altered by PEG contents
Greg C Randall et al.
Lab on a chip, 6(4), 516-525 (2006-03-31)
The ability to controllably and continuously stretch large DNA molecules in a microfluidic format is important for gene mapping technologies such as Direct Linear Analysis (DLA). We have recently shown that electric field gradients can be readily generated in a
Stress Relaxation by Addition- Fragmentation Chain Transfer in Highly Cross-Linked Thiol- Yne Networks
Park HY, et al.
Macromolecules, 43(24), 10188-10190 (2010)
Wei Yu et al.
Materials (Basel, Switzerland), 12(23) (2019-12-01)
Polymeric microparticles were produced following a three-step procedure involving (i) the production of an aqueous nanoemulsion of tri and monofunctional acrylate-based monomers droplets by an elongational-flow microemulsifier, (ii) the production of a nanosuspension upon the continuous-flow UV-initiated miniemulsion polymerization of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service