Skip to Content
Merck
All Photos(3)

Key Documents

415952

Sigma-Aldrich

Diphenyl(2,4,6-trimethylbenzoyl)phosphine oxide

97%

Synonym(s):

(2,4,6-Trimethylbenzoyl)diphenylphosphine oxide, (Diphenylphosphoryl)(mesityl)methanone, 2,4,6-Trimethylbenzoylphenyl phosphinate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3C6H2COP(O)(C6H5)2
CAS Number:
Molecular Weight:
348.37
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

97%

form

powder

mp

88-92 °C (lit.)

SMILES string

Cc1cc(C)c(c(C)c1)C(=O)P(=O)(c2ccccc2)c3ccccc3

InChI

1S/C22H21O2P/c1-16-14-17(2)21(18(3)15-16)22(23)25(24,19-10-6-4-7-11-19)20-12-8-5-9-13-20/h4-15H,1-3H3

InChI key

VFHVQBAGLAREND-UHFFFAOYSA-N

General description

Diphenyl(2,4,6-trimethylbenzoyl)phosphine oxide (TPO) is a monoacylphosphine oxide based photoinitiator that can be incorporated in a variety of polymeric matrixes for efficient curing and color stability of the resin.[1]

Application

TPO can be used in the photo-crosslinking of PMMA composite, which can further be used as a gate insulator in organic thin film transistors (OTFTs).[2] It can also be used in the formation of UV curable urethane-acrylate coatings.[3] It may also be used in the photoinduced reaction for the formation of organophosphine compounds, which potentially find their usage as ligands with metal catalysts and reagents.[4]

Storage and Stability

light sensitive

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Repr. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PMMA-based patternable gate insulators for organic thin-film transistors
Kim TG, et al.
Synthetic Metals, 159(7-8), 749-753 (2009)
Monomer-to-polymer conversion and micro-tensile bond strength to dentine of experimental and commercial adhesives containing diphenyl (2, 4, 6-trimethylbenzoyl) phosphine oxide or a camphorquinone/amine photo-initiator system
Miletic V, et al.
Journal of Dentistry, 41(10), 918-926 (2013)
Andrew V Healy et al.
Pharmaceutics, 11(12) (2019-12-11)
The introduction of three-dimensional printing (3DP) has created exciting possibilities for the fabrication of dosage forms, paving the way for personalized medicine. In this study, oral dosage forms of two drug concentrations, namely 2.50% and 5.00%, were fabricated via stereolithography
Ela Sachyani Keneth et al.
Polymers, 12(3) (2020-03-27)
Shape memory polymers are attractive smart materials that have many practical applications and academic interest. Three-dimensional (3D) printable shape memory polymers are of great importance for the fabrication of soft robotic devices due to their ability to build complex 3D
I Capasso et al.
Scientific reports, 10(1), 612-612 (2020-01-19)
Motivated by the hierarchical micro and nanoscale features in terms of porosity of diatomite, the production of ceramic-graded porous foams with tailored porosity, obtained by using it as raw material, has been proposed. The main challenge during the foam-production process

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service