Skip to Content
Merck
All Photos(2)

Key Documents

396281

Sigma-Aldrich

2,4,6,8-Tetramethyl-2,4,6,8-tetravinylcyclotetrasiloxane

Synonym(s):

2,4,6,8-Tetraethenyl-2,4,6,8-tetramethylcyclotetrasiloxane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-Si(CH3)(CH=CH2)O-]4
CAS Number:
Molecular Weight:
344.66
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

liquid

Quality Level

refractive index

n20/D 1.434 (lit.)

bp

111-112 °C/10 mmHg (lit.)

mp

−44 °C (dec.) (lit.)

density

0.997 g/mL at 25 °C (lit.)

SMILES string

C[Si]1(O[Si](C)(O[Si](C)(O[Si](C)(O1)C=C)C=C)C=C)C=C

InChI

1S/C12H24O4Si4/c1-9-17(5)13-18(6,10-2)15-20(8,12-4)16-19(7,11-3)14-17/h9-12H,1-4H2,5-8H3

InChI key

VMAWODUEPLAHOE-UHFFFAOYSA-N

General description

In the presence of a Pd catalyst, this reagent cross-couples with aryl bromides to give styrene derivatives.

Application

  • Cyclic siloxanes conjugated with fluorescent aromatic compounds as fluoride sensors: This study explores the use of cyclic siloxanes conjugated with aromatic compounds for sensing applications, offering a pathway for academic chemists interested in sensor technology and fluorescence studies (N Prigyai et al., 2020).
  • Synthesis, film morphology and performance of novel crosslinked polysiloxane with end-capped epoxy groups on cotton substrates: This article examines the chemical modification of cotton substrates using polysiloxane derivatives, which can be crucial for developing advanced textile coatings (L Hao et al., 2014).
  • Stabilizing Li-rich layered oxide cathode interface by using silicon-based electrolyte additive: This research investigates the use of a silicon-based additive derived from 2,4,6,8-tetramethyl-2,4,6,8-tetravinylcyclotetrasiloxane to enhance the stability of lithium-rich cathodes, important for chemists working with energy storage technologies (T Huang et al., 2024).

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohand O Saed et al.
Scientific reports, 10(1), 6609-6609 (2020-04-22)
Liquid crystalline elastomers (LCE) undergo reversible shape changes in response to stimuli, which enables a wide range of smart applications, in soft robotics, adhesive systems or biomedical medical devices. In this study, we introduce a new dynamic covalent chemistry based on
Ashish Pandey et al.
Soft matter, 15(13), 2897-2904 (2019-03-09)
Nanoimprinting with rigid molds offers almost unlimited pattern resolution, but it suffers from high sensitivity to defects, and is limited to pattering flat surfaces. These limitations can be addressed by nanoimprinting with soft molds. However, soft molds have been used
Byoung Choul Kim et al.
Small (Weinheim an der Bergstrasse, Germany), 10(19), 4020-4029 (2014-06-20)
Adjustable fluidic structures play an important role in microfluidic systems. Fracture of multilayered materials under applied tension has been previously demonstrated as a convenient, simple, and inexpensive approach to fabricate nanoscale adjustable structures; here, it is demonstrated how to extend
A Paul et al.
Soft matter, 13(34), 5665-5675 (2017-07-25)
We show that submicron-sized patterns can be imprinted into soft, recombinant-engineered protein hydrogels (here elastin-like proteins, ELP) by transferring wavy patterns from polydimethylsiloxane (PDMS) molds. The high-precision topographical tunability of the relatively stiff PDMS is translated to a bio-responsive, soft
Alberto Portone et al.
Scientific reports, 9(1), 7370-7370 (2019-05-16)
Polythiophenes are the most widely utilized semiconducting polymers in organic electronics, but they are scarcely exploited in photonics due to their high photo-induced absorption caused by interchain polaron pairs, which prevents the establishment of a window of net optical gain.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service