Skip to Content
Merck
All Photos(1)

Documents

439231

Sigma-Aldrich

Hexadecyltrimethylammonium hydroxide solution

10 wt. % in H2O

Synonym(s):

N,N,N-Trimethylhexadecan-1-aminium hydroxide, Cetyltrimethylammonium hydroxide, Trimethylhexadecylammonium hydroxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15N(OH)(CH3)3
CAS Number:
Molecular Weight:
301.55
Beilstein:
3723055
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

form

liquid

concentration

10 wt. % in H2O

refractive index

n20/D 1.351

density

0.988 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

[OH-].CCCCCCCCCCCCCCCC[N+](C)(C)C

InChI

1S/C19H42N.H2O/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(2,3)4;/h5-19H2,1-4H3;1H2/q+1;/p-1

InChI key

WJLUBOLDZCQZEV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Hexadecyltrimethylammonium hydroxide solution is a surfactant used in:
  • Synthesis of microporous and mesoporous aluminosilicate molecular sieves such as MCM-41 and MCM-48.
  • Hydrothermal preparation of manganese dioxide (MnO2) nanoflower.
  • Synthesis of cetyltrimethylammonium hydroperoxide, a reagent used for promoting phosphate ester hydrolysis.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cetyltrimethylammonium hydroperoxide: an efficient reagent for promoting phosphate ester hydrolysis.
Toullec, Jean and Moukawim, Mohamed
Chemical Communications (Cambridge, England), 434(2), 221-222 (1996)
Hydrothermal preparation and the capacitance of hierarchical MnO2 nanoflower.
Zhu, Gang et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 434(27), 42-48 (2013)
Synthesis of macrostructured MCM-48 molecular sieves.
Danumah, Ch et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 44(27), 241-247 (2001)
Ordered mesoporous molecular sieves synthesized by a liquid-crystal template mechanism.
Kresge C T, et al.
Nature, 359(6397), 710-710 (1992)
A new family of mesoporous molecular sieves prepared with liquid crystal templates.
Beck JS
Journal of the American Chemical Society, 114(27), 10834-10843 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service