Skip to Content
Merck
All Photos(1)

Documents

35915

Sigma-Aldrich

Sodium dichloroisocyanurate dihydrate

98.0% (AT)

Synonym(s):

NaDCC, Dichloroisocyanuric acid sodium salt dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3Cl2N3NaO3 · 2H2O
CAS Number:
Molecular Weight:
255.98
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.22

Assay

98.0% (AT)

reaction suitability

reagent type: oxidant

impurities

~0.5% free chloride

SMILES string

[Na+].[H]O[H].[H]O[H].[O-]C1=NC(=O)N(Cl)C(=O)N1Cl

InChI

1S/C3HCl2N3O3.Na.2H2O/c4-7-1(9)6-2(10)8(5)3(7)11;;;/h(H,6,9,10);;2*1H2/q;+1;;/p-1

InChI key

PYILKOIEIHHYGD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium dichloroisocyanurate dihydrate is a solid chlorine bleaching agent generally used as an alternative to sodium hypochlorite for water treatment.

Application

Calibration substance for the photometric determination of active chlorine. Guidelines for the analysis of waste water and surface water, Part 1; 28/3; Federal Department of Home Affairs 1982.
Sodium dichloroisocyanurate dihydrate can be used as a chlorine source to convert alkoxyl-s-triazines into N-chlorinated alkoxy-s-triazines.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andreja Škiljaica et al.
Plant molecular biology, 102(4-5), 359-372 (2019-12-19)
Protein degradation is essential in plant growth and development. The stability of Cullin3 substrate adaptor protein BPM1 is regulated by multiple environmental cues pointing on manifold control of targeted protein degradation. A small family of six MATH-BTB genes (BPM1-6) is
N-chloro-alkoxy-s-triazine-based antimicrobial additives: preparation, characterization, and antimicrobial and biofilm-controlling functions.
Sun X, et al.
Industrial & Engineering Chemistry Research, 48(2), 607-612 (2008)
I H Rweyendela et al.
SADJ : journal of the South African Dental Association = tydskrif van die Suid-Afrikaanse Tandheelkundige Vereniging, 64(5), 208-208 (2009-09-04)
Irreversible hydrocolloid (alginate) impressions are dimensionally unstable and difficult to disinfect. To evaluate the antimicrobial efficacy of a chlorite disinfectant (Presept) and a new formulation chlorine dioxide based disinfectant (Aseptrol) on irreversible hydrocolloid (alginate) impression material. Alginate blocks were contaminated
Daniele S Lantagne et al.
The American journal of tropical medicine and hygiene, 83(1), 135-143 (2010-07-03)
Almost a billion persons lack access to improved drinking water, and diarrheal diseases cause an estimated 1.87 million deaths per year. Sodium dichloroisocyanurate (NaDCC) tablets are widely recommended for household water treatment to reduce diarrhea. Because NaDCC is directly added
Bleaching agents.
Kirk-Othmer Encyclopedia of Chemical Technology (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service