Skip to Content
Merck
All Photos(3)

Key Documents

C87908

Sigma-Aldrich

Cyanamide solution

50 wt. % in H2O

Synonym(s):

Alzogur, Amidocyanogen, Amino cyanide, Carbamonitrile, Carbimide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCNH2
CAS Number:
Molecular Weight:
42.04
Beilstein:
1732569
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

concentration

50 wt. % in H2O

refractive index

n20/D 1.405

density

1.082 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

NC#N

InChI

1S/CH2N2/c2-1-3/h2H2

InChI key

XZMCDFZZKTWFGF-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Chronic 3 - Carc. 2 - Eye Dam. 1 - Repr. 2 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2

Target Organs

Thyroid

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuan Liu et al.
Nanomaterials (Basel, Switzerland), 9(6) (2019-06-05)
To improve the power generation of a microbial fuel cell (MFC), a porous nitrogen-doped graphene/carbon black (NG/CB) composite as efficient oxygen reduction reaction (ORR) electrocatalyst was successfully synthesized by pyrolyzing graphene oxide (GO) encapsulated CB with cetyltrimethyl ammonium bromide as
Santiago Signorelli et al.
Journal of experimental botany, 71(2), 719-729 (2019-05-01)
The physiological constraints on bud burst in woody perennials, including vascular development and oxygenation, remain unresolved. Both light and tissue oxygen status have emerged as important cues for vascular development in other systems; however, grapevine buds have only a facultative
Ricardo Vergara et al.
Plant molecular biology, 79(1-2), 171-178 (2012-04-03)
It has been reported that dormancy-breaking compound hydrogen cyanamide (HC) stimulates the fermentative pathway and inhibits respiration in grapevine-buds, suggesting in this way, that a respiratory stress must be involved in the release of buds from dormancy. Here, we tested
Carmen Harb et al.
Inorganic chemistry, 52(3), 1621-1630 (2013-01-24)
Nine [Ru(Tp)(dppe)L] complexes, where Tp is hydrotris(pyrazol-1-yl)borate, dppe is ethylenebis(diphenylphosphine), and L is (4-nitrophenyl)cyanamide (NO(2)pcyd(-)), (2-chlorophenyl)cyanamide (2-Clpcyd(-)), (3-chlorophenyl)cyanamide (3-Clpcyd(-)), (2,4-dichlorophenyl)cyanamide (2,4-Cl(2)pcyd(-)), (2,3-dichlorophenyl)cyanamide (2,3-Cl(2)pcyd(-)), (2,5-dichlorophenyl)cyanamide (2,5-Cl(2)pcyd(-)), (2,4,5-trichlorophenyl)cyanamide (2,4,5-Cl(3)pcyd(-)), (2,3,5,6-tetrachlorophenyl)cyanamide (2,3,5,6-Cl(4)pcyd(-)), and (pentachlorophenyl)cyanamide (Cl(5)pcyd(-)), and the dinuclear complex [{Ru(Tp)(dppe)}(2)(μ-adpc)], where adpc(2-) is
Dorota Soltys et al.
Planta, 236(5), 1629-1638 (2012-08-01)
Cyanamide (CA) has been reported as a natural compound produced by hairy vetch (Vicia villosa Roth.) and it was shown also to be an allelochemical, responsible for strong allelopathic potential in this species. CA phytotoxicity has been demonstrated on various

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service