Skip to Content
Merck
All Photos(1)

Documents

246034

Sigma-Aldrich

Benzyltrimethylammonium hydroxide solution

40 wt. % in H2O

Synonym(s):

N,N,N-Trimethyl-N-benzylammonium hydroxide, N,N,N-trimethyl-1-phenylmethanaminium hydroxide, N,N,N-trimethylbenzenemethanaminium hydroxide, Triton B

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2N(OH)(CH3)3
CAS Number:
Molecular Weight:
167.25
Beilstein:
3917256
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

concentration

40 wt. % in H2O

refractive index

n20/D 1.43

density

1.059 g/mL at 25 °C

SMILES string

[OH-].C[N+](C)(C)Cc1ccccc1

InChI

1S/C10H16N.H2O/c1-11(2,3)9-10-7-5-4-6-8-10;/h4-8H,9H2,1-3H3;1H2/q+1;/p-1

InChI key

NDKBVBUGCNGSJJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Benzyltrimethylammonium hydroxide solution is used as a phase transfer catalyst. It can assist in situ transesterificaion to synthesize biodiesel.

Application

Benzyltrimethylammonium hydroxide can be used:
  • As a base in the synthesis of activated ynesulfonamides and tertiary enesulfonamides.
  • In the synthesis of 3-indolyl-3-hydroxy oxindoles by treating isatin with indole.
  • As a precursor for the preparation of quaternary ammonium acetate, benzyltrimethylammonium acetate (NBz111AcO). NBz111AcO solution in DMSO can be used to dissolve cellulose.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metal-Free Synthesis of Activated Ynesulfonamides and Teritary Enesulfonamides
Andna L and Miesch L
Organic & Biomolecular Chemistry (2019)
Dependence of cellulose dissolution in quaternary ammonium-based ionic liquids/DMSO on the molecular structure of the electrolyte
Kostag M and El Seoud OA
Carbohydrate Polymers, 205(1), 524-532 (2019)
Parametric study and optimization of in situ transesterification of Jatropha curcas L assisted by benzyltrimethylammonium hydroxide as a phase transfer catalyst via response surface methodology
Hailegiorgis SM, et al.
Biomass and Bioenergy, 63-73}-63-73} (2013)
Triton B-Assisted, Efficient, and Convenient Synthesis of 3-Indolyl-3-hydroxy Oxindoles in Aqueous Medium
Meshram HM, et al.
Synthetic Communications, 40(1), 39-45 (2009)
L Sangeetha Vedula et al.
Archives of biochemistry and biophysics, 466(2), 260-266 (2007-08-07)
Trichodiene synthase is a terpenoid cyclase that catalyzes the cyclization of farnesyl diphosphate (FPP) to form the bicyclic sesquiterpene hydrocarbon trichodiene (89%), at least five sesquiterpene side products (11%), and inorganic pyrophosphate (PP(i)). Incubation of trichodiene synthase with 2-fluorofarnesyl diphosphate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service