Skip to Content
Merck
All Photos(1)

Key Documents

187364

Sigma-Aldrich

Cyanamide

99%

Synonym(s):

Carbimide, Hydrogen cyanamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCNH2
CAS Number:
Molecular Weight:
42.04
Beilstein:
1732569
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
38060506
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

crystals

contains

stabilizer

bp

83 °C/0.5 mmHg (lit.)

mp

45-46 °C (lit.)

solubility

ethanol: soluble 10%, clear to hazy, colorless to faintly yellow
water: soluble

functional group

amine

storage temp.

2-8°C

SMILES string

NC#N

InChI

1S/CH2N2/c2-1-3/h2H2

InChI key

XZMCDFZZKTWFGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Cyanamide is an effective agent to treat alcoholism. Cyanamide in aqueous solution or in the form of its calcium salt is used as a fertilizer in agriculture. It is a potent inhibitor of liver aldehyde dehydrogenases in vivo.

Application

Cyanamide was used as nitrogen source in thermal transformation of metal oxide nanoparticles into nanocrystalline metal nitrides. It was used in the synthesis of mesoporous graphitic C3N4.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Aquatic Chronic 3 - Carc. 2 - Eye Dam. 1 - Repr. 2 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2

Target Organs

Thyroid

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anand K Ganesan et al.
PLoS genetics, 4(12), e1000298-e1000298 (2008-12-06)
Melanin protects the skin and eyes from the harmful effects of UV irradiation, protects neural cells from toxic insults, and is required for sound conduction in the inner ear. Aberrant regulation of melanogenesis underlies skin disorders (melasma and vitiligo), neurologic
Woong Kwon et al.
Polymers, 12(7) (2020-07-08)
This study investigates the detoxification properties of guanidinylated chitosan against chemical warfare agents and its application to the preparation of military protective clothing. Guanidinylated chitosan was synthesized by chitosan guanidinylation with cyanamide. The detoxification properties of the guanidinylated chitosan were
Inhibition of aldehyde dehydrogenase in brain and liver by cyanamide.
R A Deitrich et al.
Biochemical pharmacology, 25(24), 2733-2737 (1976-12-15)
U H Maier-Greiner et al.
Proceedings of the National Academy of Sciences of the United States of America, 88(10), 4260-4264 (1991-05-15)
A protein was purified from crude extracts of the soil fungus Myrothecium verrucaria by gel filtration and hydrophobic chromatography to homogeneity; this protein catalyzed the stoichiometric hydration of the fertilizer cyanamide to urea with high substrate specificity. This cyanamide hydratase
Mesoporous graphitic carbon nitride as a versatile, metal-free catalyst for the cyclisation of functional nitriles and alkynes.
Goettmann F, et al.
New. J. Chem., 31(8), 1455-1460 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service