Skip to Content
Merck
All Photos(1)

Documents

392111

Sigma-Aldrich

Isobornyl methacrylate

technical grade

Synonym(s):

IBMA, Methacrylic acid isobornyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22O2
CAS Number:
Molecular Weight:
222.32
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

form

liquid

contains

150 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.477 (lit.)

bp

127-129 °C/15 mmHg (lit.)

density

0.983 g/mL at 25 °C (lit.)

SMILES string

CC(=C)C(=O)OC1C[C@H]2CC[C@]1(C)C2(C)C

InChI

1S/C14H22O2/c1-9(2)12(15)16-11-8-10-6-7-14(11,5)13(10,3)4/h10-11H,1,6-8H2,2-5H3/t10-,11?,14+/m1/s1

InChI key

IAXXETNIOYFMLW-JENJKZFGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isobornyl methacrylate is a reactive solvent with low vapor pressure. It facilitates the free radical polymerization by forming a polymer with high glass transition temperature (Tg).

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

237.2 °F - Pensky-Martens closed cup

Flash Point(C)

114 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amid Ranjkesh et al.
ACS applied materials & interfaces, 11(29), 26314-26322 (2019-06-28)
Recently, single-substrate flexible liquid crystal (LC) devices have attracted considerable attention because they can provide desirable shapes, small weight, flexibility, and rollability. In this work, we fabricate a flexible single-substrate thermoresponsive cholesteric LC (CLC) film by a facile method called
Surface energies of linear and cross-linked polymers based on isobornyl methacrylate and methacryl-heptaisobutyl POSS
Zucchi IA, et al.
European Polymer Journal, 45(2), 325-331 (2009)
Isobornyl methacrylate as a reactive solvent of polyethylene
Schnell M, et al.
Macromolecular Materials and Engineering, 289(7), 642-647 (2004)
Dylan G Gee et al.
Human brain mapping, 36(7), 2558-2579 (2015-03-31)
Multisite neuroimaging studies can facilitate the investigation of brain-related changes in many contexts, including patient groups that are relatively rare in the general population. Though multisite studies have characterized the reliability of brain activation during working memory and motor functional
S Wünsch et al.
The Journal of cell biology, 140(2), 335-345 (1998-02-28)
Here we describe the identification and characterization of a physiological marker that is associated with the chloroquine-resistant (CQR) phenotype in the human malarial parasite Plasmodium falciparum. Single cell in vivo pH measurements revealed that CQR parasites consistently have an elevated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service