Skip to Content
Merck
All Photos(1)

Key Documents

268542

Sigma-Aldrich

Hydroxypropyl methacrylate

Mixture of hydroxypropyl and hydroxyisopropyl methacrylates, 97%, contains 180-220 ppm monomethyl ether hydroquinone as inhibitor

Synonym(s):

HPMA

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
1752228
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

>1 (vs air)

vapor pressure

0.05 mmHg ( 20 °C)

Assay

97%

form

liquid

contains

180-220 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.447 (lit.)

bp

57 °C/0.5 mmHg (lit.)

density

1.066 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=C)C(=O)OCCCO

InChI

1S/C7H12O3/c1-5(2)7(9)10-6(3)4-8/h6,8H,1,4H2,2-3H3

InChI key

ZMARGGQEAJXRFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Synthesis of poly hydroxypropyl methacrylate cryogel incorporated with Zn/Ce substituted hydroxyapatite nanoparticles for rejuvenation of femoral fracture: This study explores the synthesis of a cryogel incorporating hydroxypropyl methacrylate and Zn/Ce substituted hydroxyapatite nanoparticles, aimed at bone fracture healing (H Zhou, H Jiao, J Xu, Y Liu, S Wei, 2019).

  • Poly(Hydroxypropyl methacrylate-co-glycidyl methacrylate): Facile synthesis of well-defined hydrophobic gels containing hydroxy-functional methacrylates: This article presents the synthesis of hydrophobic gels using hydroxypropyl methacrylate and glycidyl methacrylate, useful in various material applications (N Orakdogen, B Sanay, 2017).

  • Initiated chemical vapor deposition of poly(Hydroxypropyl methacrylate) thin films: The study covers the chemical vapor deposition process for creating thin films of poly(hydroxypropyl methacrylate) on membranes, enhancing their functional properties (E Sevgili, M Karaman, 2019).

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Muta. 1B - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

203.0 °F - closed cup

Flash Point(C)

95 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomáš Etrych et al.
Journal of controlled release : official journal of the Controlled Release Society, 154(3), 241-248 (2011-06-28)
Herein, new biodegradable star polymer-doxorubicin conjugates designed for passive tumor targeting were investigated, and their synthesis, physico-chemical characterization, drug release, biodegradation, biodistribution and in vivo anti-tumor efficacy are described. In the conjugates, the core formed by poly(amidoamine) (PAMAM) dendrimers was
Qingyu Xiang et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 80(2), 379-386 (2011-11-22)
In several groups of malignant tumors including head and neck tumors, a protein named Hsp47/CBP2 leaked from the cell was expressed on the tumor cell surface. Several synthetic peptides have been identified as effective ligands for binding to Hsp47/CBP2. This
Russell N Johnson et al.
Biomacromolecules, 13(3), 727-735 (2012-02-01)
High-molecular-weight, branched N-(2-hydroxypropyl)methacrylamide (HPMA) copolymers were synthesized and conjugated with Fab' fragments of the anti-CD20 antibody, 1F5. This produced multivalent conjugates with varying valency (amount of Fab' per macromolecule) targeted to the B-cell antigen CD20. The apoptotic activity of the
Adam J Gormley et al.
Journal of controlled release : official journal of the Controlled Release Society, 166(2), 130-138 (2012-12-25)
Effective drug delivery to tumors requires both transport through the vasculature and tumor interstitium. Previously, it was shown that gold nanorod (GNR) mediated plasmonic photothermal therapy (PPTT) is capable of increasing the overall accumulation of N-(2-hydroxypropyl)methacrylamide (HPMA) copolymers in prostate
Annette Kelsch et al.
Biomacromolecules, 13(12), 4179-4187 (2012-11-28)
In this work we describe the application of amphiphilic N-(2-hydroxypropyl)methacrylamide (HPMA)-based copolymers as polymeric surfactants in miniemulsion techniques. HPMA-based copolymers with different ratios of HPMA (hydrophilic) to laurylmethacrylate (LMA; hydrophobic) units were synthesized by RAFT polymerization and postpolymerization modification. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service