Skip to Content
Merck
All Photos(1)

Key Documents

128635

Sigma-Aldrich

2-Hydroxyethyl methacrylate

contains ≤250 ppm monomethyl ether hydroquinone as inhibitor, 97%

Synonym(s):

1,2-Ethanediol mono(2-methylpropenoate), Glycol methacrylate, HEMA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=C(CH3)COOCH2CH2OH
CAS Number:
Molecular Weight:
130.14
Beilstein:
1071583
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

5 (vs air)

vapor pressure

0.01 mmHg ( 25 °C)

Assay

97%

form

liquid

contains

≤250 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.453 (lit.)

bp

67 °C/3.5 mmHg (lit.)

density

1.073 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=C)C(=O)OCCO

InChI

1S/C6H10O3/c1-5(2)6(8)9-4-3-7/h7H,1,3-4H2,2H3

InChI key

WOBHKFSMXKNTIM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Hydroxyethyl methacrylate (HEMA) was the first monomer to be used to synthesize hydrogels for biomedical applications.The water swelling properties of HEMA are enhanced by copolymerization with more hydrophilic monomers.HEMA finds wide applications for drug delivery.

Application

HEMA is used in the synthesis of biologically functional poly (2-hydroxyethyl methacrylate) (PHEMA) coplymers. It is also used to prepare light responsive membranes of PHEMA, HEMA/ acrylamide based specific drug release hydrogel, and water soluble HEMA/methacrylic acid hydrogel for drug delivery. Effects of HEMA on the migration of dental pulp stem cells (in vitro) may be studied.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

222.8 °F - closed cup

Flash Point(C)

106 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giovanna C R M Schver et al.
International journal of pharmaceutics, 581, 119292-119292 (2020-04-04)
The use of water-insoluble carriers for amorphous solid dispersions (ASDs) has attracted more recent interest as the kinetic solubility profiles (KSP) from these systems can achieve a more sustained level of supersaturation when compared with ASDs based on water-soluble polymers.
Drake W Williams et al.
Journal of endodontics, 39(9), 1156-1160 (2013-08-21)
Cell migration is an important step in pulpal wound healing. Although components in the resin-based dental materials are known to have adverse effects on pulp wound healing including proliferation and mineralization, their effects on cell migration have been scarcely examined.
Influence of the ionic character of a drug on its release rate from hydrogels based on 2-hydroxyethylmethacrylate and acrylamide synthesized by photopolymerization
Gomez ML, et al.
Express Polymer Letters, 6(30, 189-197 (2012)
Preparation of Light-responsive Membranes by a Combined Surface Grafting and Postmodification Process.
Scholler K, et al.
Journal of Visualized Experiments, 85, e51680-e51680 (2014)
Thomas J Dursch et al.
Biomaterials, 35(2), 620-629 (2013-10-24)
Two-photon confocal microscopy and back extraction with UV/Vis-absorption spectrophotometry quantify equilibrium partition coefficients, k, for six prototypical drugs in five soft-contact-lens-material hydrogels over a range of water contents from 40 to 92%. Partition coefficients were obtained for acetazolamide, caffeine, hydrocortisone

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service