Skip to Content
Merck
All Photos(2)

Key Documents

346683

Sigma-Aldrich

2-Phenoxyaniline

95%

Synonym(s):

2-Aminophenyl phenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OC6H4NH2
CAS Number:
Molecular Weight:
185.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

bp

170 °C/18 mmHg (lit.)

mp

47-49 °C (lit.)

functional group

phenoxy

SMILES string

Nc1ccccc1Oc2ccccc2

InChI

1S/C12H11NO/c13-11-8-4-5-9-12(11)14-10-6-2-1-3-7-10/h1-9H,13H2

InChI key

NMFFUUFPJJOWHK-UHFFFAOYSA-N

General description

2-Phenoxyaniline forms crystalline 2:1 (host-to-guest) inclusion complexes with β-cyclodextrin.

Application

2-Phenoxyaniline may be used in the preparation of sodium primary amide complex, 2-PhOC6H4NHNa·NMe[(CH2)2NMe2]2. It was used in the preparation of 2-acetoaminodiphenyl ether. It may be used in the preparation of 4-methyl-2-(2-phenoxyphenyl)azo-phenol.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susana S Braga et al.
Organic & biomolecular chemistry, 1(5), 873-878 (2003-08-22)
Crystalline 1:1 inclusion complexes with beta-cyclodextrin (beta-CD) and the sodium salt of nimesulide (4-nitro-2-phenoxymethanesulfonanilide), and the sodium salt of the derivative 2-phenoxymethanesulfonanilide, have been prepared by co-precipitation from aqueous solution. The presence of true inclusion complexes was supported by elemental
Synthesis and Pharmacological Evaluation of Certain Schiff Bases and Thiazoldine derivatives as AT1 angiotension-II (AII) receptor antagonists.
Shreenivas MT, et al.
Journal of Pharmaceutical Science and Technology, 1(2), 88-94 (2009)
Ferenc Szurdoki et al.
Journal of agricultural and food chemistry, 50(1), 29-40 (2002-01-05)
Sensitive and selective enzyme-linked immunosorbent assays (ELISAs) in the immobilized antigen format were developed for fenoxycarb (1), an insect growth regulator (IGR). The parent molecule [ethyl 2-(4-phenoxyphenoxy)ethylcarbamate] was derivatized at several positions to obtain haptens (2-5) that were used to
The first structurally characterised sodium primary amide complex; synthesis and crystal structure of 2-PhOC6H4NHNa? NMe [(CH2) 2NMe2] 2.
Cragg-Hine I, et al.
J. Chem. Soc., Dalton Trans., 19, 2901-2902 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service