Skip to Content
Merck
All Photos(3)

Documents

148075

Sigma-Aldrich

Camphor

96%

Synonym(s):

(±)-Camphor, 1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
Beilstein:
1907611
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

5.2 (vs air)

vapor pressure

4 mmHg ( 70 °C)

Assay

96%

autoignition temp.

870 °F

expl. lim.

3.5 %

bp

204 °C (lit.)

mp

175-177 °C (lit.)

solubility

ethanol: soluble 1 g/L, clear, colorless

SMILES string

[H][C@](CC1=O)(CC2)C(C)(C)[C@]12C

InChI

1S/C10H16O/c1-9(2)7-4-5-10(9,3)8(11)6-7/h7H,4-6H2,1-3H3/t7-,10+/m1/s1

InChI key

DSSYKIVIOFKYAU-XCBNKYQSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Camphor is a novel carbon nanotube precursor.

Application

Camphor was used in the synthesis of single-walled nanotubes by chemical vapor deposition. It was used in two-phase based hollow fibre liquid-phase microextraction procedure for migration analysis of food packagings containing essential oils.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 2 Inhalation

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

147.9 °F - closed cup

Flash Point(C)

64.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Davinson Pezo et al.
Journal of chromatography. A, 1174(1-2), 85-94 (2007-09-04)
A two-phase based hollow fibre liquid-phase microextraction (HFLPME) with a high automatization degree and able to process up to six samples simultaneously by means of a multiple channel syringe pump has been successfully developed. The experimental set-up allows to carry
A simple method of producing aligned carbon nanotubes from an unconventional precursor-Camphor.
Kumar M and Ando Y.
Chemical Physics Letters, 374(5), 521-526 (2003)
Mechanism of carbon nanotube growth from camphor and camphor analogs by chemical vapor deposition.
Andrews RJ, et al.
Carbon, 44(2), 341-347 (2006)
Simon P Skinner et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(29), 9022-9027 (2015-07-02)
The energy landscapes of proteins are highly complex and can be influenced by changes in physical and chemical conditions under which the protein is studied. The redox enzyme cytochrome P450cam undergoes a multistep catalytic cycle wherein two electrons are transferred
Tamera J Corte et al.
American journal of respiratory and critical care medicine, 190(2), 208-217 (2014-06-18)
Pulmonary hypertension (PH) associated with fibrotic idiopathic interstitial pneumonia (IIP; idiopathic pulmonary fibrosis and nonspecific interstitial pneumonia) confers important additional morbidity and mortality. To evaluate the safety and clinical efficacy of the dual endothelin-1 receptor antagonist bosentan in this patient

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service