Skip to Content
Merck
All Photos(8)

Key Documents

D80002

Sigma-Aldrich

DCC

99%

Synonym(s):

N,N′-Dicyclohexylcarbodiimide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11N=C=NC6H11
CAS Number:
Molecular Weight:
206.33
Beilstein:
610662
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

reaction suitability

reaction type: Coupling Reactions

bp

122-124 °C/6 mmHg (lit.)

mp

34-35 °C (lit.)

SMILES string

C1CCC(CC1)N=C=NC2CCCCC2

InChI

1S/C13H22N2/c1-3-7-12(8-4-1)14-11-15-13-9-5-2-6-10-13/h12-13H,1-10H2

InChI key

QOSSAOTZNIDXMA-UHFFFAOYSA-N

Gene Information

human ... EPHX2(2053)
mouse ... Ephx2(13850)

Looking for similar products? Visit Product Comparison Guide

General description

DCC is the acronym for N,N′-dicyclohexylcarbodiimide. In peptide synthesis, it is utilized for the activation of carboxyl group. The structure of DCC has been analyzed based on IR and Raman spectral data. It mediates the esterification of poly(vinyl alcohol) with free levulinic acid to form poly(vinyl alcohol-co-vinyl levulinate).

Application

Carboxy group activating reagent for peptide synthesis.
DCC has been used in the preparation of titanate nanotubes-phthalocyanine(TiONts-Pc) nanohybrids and 7-((tert-butyldimethylsilyl)oxy)hepta-2,4-diyn-1-yl propynoate.
DCC may be used to promote the esterification of
7-((tert-butyldimethylsilyl)oxy)hepta-2,4-diyn-1-ol with propiolic acid to form 7-((tert-butyldimethylsilyl)oxy)hepta-2,4-diyn-1-yl propynoate.
It can also used to synthesize:
  • 1,3-Thiazetedine derivatives via [2+2] cycloaddition with 2-phenylethenyl- and 2-(4-nitrophenyl)ethenyl isothiocyanates.
  • 1,3,5-Oxadiazine-4-thiones via [4+2] cycloaddition with benzoyl isothiocyanates.
  • Sterically hindered 1,3,4-oxadiazole derivatives by reacting with (N-isocyanimino)triphenylphosphorane in the presence of aromatic (or heteroaromatic) carboxylic acids.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vibrational spectroscopy and DFT calculations of N,N'-dicyclohexylcarbodiimide.
Chowdhry BZ, et al.
Journal of Raman Spectroscopy, 42(2), 230-238 (2011)
Phthalocyanine-titanate nanotubes: a promising nanocarrier detectable by optical imaging in the so-called imaging window.
Paris J, et al.
Royal Society of Chemistry Advances, 5(9), 6315-6322 (2015)
Jones, J.
The Chemical Synthesis of Peptides, International Series of Monographs on Chemistry, 23 (1994)
Svenja Bockelmann et al.
The Journal of biological chemistry, 285(49), 38304-38314 (2010-10-05)
The macrolactone archazolid is a novel, highly specific V-ATPase inhibitor with an IC(50) value in the low nanomolar range. The binding site of archazolid is presumed to overlap with the binding site of the established plecomacrolide V-ATPase inhibitors bafilomycin and
N,N-dicyclohexylcarbodiimide assisted synthesis and characterization of poly (vinyl alcohol-co-vinyl levulinate).
Wang, YM, et al.
Polymer, 46(23), 9793-9802 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service