Skip to Content
Merck
All Photos(3)

Key Documents

132292

Sigma-Aldrich

2-Vinylpyridine

97%

Synonym(s):

α-Vinylpyridine, 2-Ethenylpyridine, 2-Pyridylethylene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7N
CAS Number:
Molecular Weight:
105.14
Beilstein:
104505
EC Number:
MDL number:
UNSPSC Code:
12162002
eCl@ss:
39151715
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

vapor pressure

10 mmHg ( 44.5 °C)

Assay

97%

form

liquid

contains

0.1 wt. % p-tert-butylcatechol as inhibitor

refractive index

n20/D 1.549 (lit.)

bp

79-82 °C/29 mmHg (lit.)

density

0.975 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

C=Cc1ccccn1

InChI

1S/C7H7N/c1-2-7-5-3-4-6-8-7/h2-6H,1H2

InChI key

KGIGUEBEKRSTEW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Vinylpyridine (2VP) belongs to the class of monomers known as vinyl monomers. It is used as a monomer in the synthesis of various polymers such as poly(2-vinylpyridine) (P2VP), poly(2-vinylpyridine-co-styrene), poly(2-vinylpyridine-co-butadiene) etc. These polymers find applications in membrane and separation technologies, ion exchange resins, electronic devices, sensors, energy storage systems, and drug delivery systems. Additionally, 2VP polymers are used as binders in adhesives and coatings due to their excellent adhesion properties. They can provide strong bonding to various substrates.

2-Vinylpyridine (2VP) is a water soluble pyridine derivative that can be used in the synthesis of poly(2-vinylpyridine) (P2VP) by anionic polymerization. It can be industrially prepared by treatment of 2-methylpyridine with an aqueous solution of formaldehyde in a temperature range of 150-200°C.[1][2][3]

Application

2-Vinylpyridine can be used as a monomer:
  • In the preparation of P2VP oligomers to study their properties and behavior on various surfaces, with a focus on area-selective deposition techniques.
  • To synthesize pH-responsive thin film membranes, which can be used for various applications, such as separation, filtration, and catalysis.
  • To synthesize degradable block copolymer-derived nanoporous membranes, which have potential applications in various fields, including energy-related catalysis and nanoparticle synthesis.
  • To improve the effectiveness of protective polymer coatings on metals.
Additionally, 2VP can be used for a variety of applications such as dye sensitized solar cells (DSSCs) and lithium ion cells.[3][4]
Vinylation of silacyclobutanes with Ni-catalysis leading to geometrically pure (E)-silylolefins.[5]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - Skin Sens. 1

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

122.0 °F - closed cup

Flash Point(C)

50 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vasco Branco et al.
Redox biology, 13, 278-287 (2017-06-11)
Mercury (Hg) compounds target both cysteine (Cys) and selenocysteine (Sec) residues in peptides and proteins. Thus, the components of the two major cellular antioxidant systems - glutathione (GSH) and thioredoxin (Trx) systems - are likely targets for mercurials. Hg exposure
A Jurczak et al.
Redox report : communications in free radical research, 22(6), 524-533 (2017-03-16)
The aim of this study was to evaluate the role of the antioxidant barrier in the saliva of children with caries, and its impact on the colonization of cariogenic bacteria. This is a cross-sectional study of 81 children aged 1-5
Synthesis of block copolymers of 2-and 4-vinylpyridine by RAFT polymerization
Convertine AJ, et al.
Macromolecules, 36(13), 4679-4681 (2003)
Impact of 2-Vinylpyridine as Electrolyte Additive on Surface and Electrochemistry of Graphite for C/ LiMn2O4 Li-Ion Cells
Komaba S, et al.
Journal of the Electrochemical Society, 152(5), A937-A946 (2005)
Christine C Gana et al.
Biochemical pharmacology, 168, 237-248 (2019-07-16)
Members of the ABC transporter family, particularly P-glycoprotein (P-gp, ABCB1), breast cancer resistance protein (BCRP, ABCG2) and multidrug resistance protein 1 (MRP1, ABCC1) are well characterized mediators of multidrug resistance, however their pharmacological inhibition has so far failed as a

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service