Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium 3-methyl-2-oxobutyrate
Synonym(s):
α-Ketoisovaleric acid sodium salt, 2-Keto-3-methylbutyric acid sodium salt, 3-Methyl-2-oxobutanoic acid sodium salt, 3-Methyl-2-oxobutyric acid sodium salt, Ketovaline sodium salt
Linear Formula:
(CH
3
)
2
CHCOCOONa
CAS Number:
3715-29-5
Molecular Weight:
138.10
EC Number:
223-062-2
Products
Product Category
building blocks (1)
Brand
Sigma-Aldrich (1)
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
198994
Sodium 3-methyl-2-oxobutyrate,
95%
Expand
View Pricing