Skip to Content
Merck
All Photos(1)

Key Documents

190268

Sigma-Aldrich

2,6-Dibromo-4-nitroaniline

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br2C6H2(NO2)NH2
CAS Number:
Molecular Weight:
295.92
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97%

mp

205-207 °C (lit.)

functional group

bromo
nitro

SMILES string

Nc1c(Br)cc(cc1Br)[N+]([O-])=O

InChI

1S/C6H4Br2N2O2/c7-4-1-3(10(11)12)2-5(8)6(4)9/h1-2H,9H2

InChI key

YMZIFDLWYUSZCC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,6-Dibromo-4-nitroaniline has been used in the synthesis of:
  • blue disperse dyes
  • 7-bromo-5-iodo-4-oxo-1,4-dihydroquinoline-2-carboxylic acid
  • diphenols-amides such as N-(2,6-dibromo-4-nitrophenyl)-4,4-bis(4-hydroxyphenyl)-pentanamide and N-(2,6-diiodo-4-nitrophenyl)-4,4-bis(4-hydroxyphenyl)-pentanamide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polymerization by phase transfer catalysis. 24. Synthesis of condensation polymers derived from halogenated diphenol-amides with the amide group in the side chain.
Tagle LH, et al.
Polym. Bull., 42(6) (1999)
Substituent effects on the colour, dyeing and fastness properties of 4-phenylazo-1-naphthylamines and of 2-acetylamino-5-methoxy-4-N-?-cyanoethyl-N-?-hydroxyethylaminoazobenzene.
Peters AT.
Journal of the Society of Dyers and Colourists, 104(9), 344-348 (1988)
K Suresh Kumar et al.
Environmental monitoring and assessment, 127(1-3), 67-72 (2006-08-10)
Facile, selective and sensitive spectrophotometric method has been developed for the determination of bendiocarb in its insecticidal formulations, fortified water, food grains, agriculture wastewater and agriculture soil samples with prepared reagents. The method was based on alkaline hydrolysis of the
Synthesis of 7-Bromo-5-iodo-4-oxo-1, 4-dihydroquinoline-2-carboxylic acid.
Dumont F and Slegers G.
Bull. Soc. Chim. Belg., 104(8), 505-507 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service