Skip to Content
Merck
All Photos(2)

Documents

207888

Sigma-Aldrich

Bromine

reagent grade

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Br2
CAS Number:
Molecular Weight:
159.81
EC Number:
MDL number:
UNSPSC Code:
12141915
PubChem Substance ID:
NACRES:
NA.21

grade

reagent grade

Quality Level

vapor density

7.14 (vs air)

vapor pressure

175 mmHg ( 20 °C)
671 mmHg ( 55 °C)

Assay

98.0-102.0% (by Na2S2O3, titration)

form

liquid

resistivity

7.8E18 μΩ-cm, 20°C

bp

58.8 °C (lit.)

mp

−7.2 °C (lit.)

density

3.119 g/mL at 25 °C (lit.)

SMILES string

BrBr

InChI

1S/Br2/c1-2

InChI key

GDTBXPJZTBHREO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bromine is present in gaseous form in lower atmosphere generated by marine activity and as a by product produced from the leaded gasoline combustion. It is widely used as a brominating agent.

Application

Bromine may be used as one of the reactants in the preparation of:
  • quinones
  • α,β-dibromosuccinic acid
  • (Z)-ethyl 2-bromo-5-(trimethylsilyl)pent-2-en-4-ynoate
  • α,α′-dibromo-2′,4′-dinitro-2(or3 or 4)-(acetyloxy)bibenzyls
  • dibenzylnaphthols
Bromine may be used to catalyze:
  • Aziridination of olefins with chloramine-T to form N-tosyl aziridines.
  • Conversion of epoxides and CO2 to cyclic carbonates in a continuous flow system.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Metal Free Conducting PEDOS, PEDOT, and Their Analogues via an Unusual Bromine-Catalyzed Polymerization.
Patra A, et al.
Macromolecules, 48(24), 8760-8764 (2015)
Use of Bromine and Bromo-Organic Compounds in Organic Synthesis.
Saikia I, et al.
Chemical Reviews (2016)
Reaction of bromine and chlorine with phenolic compounds and natural organic matter extracts-Electrophilic aromatic substitution and oxidation.
Criquet J, et al.
Water Research, 85, 476-486 (2015)
Jennifer A Kozak et al.
Journal of the American Chemical Society, 135(49), 18497-18501 (2013-11-22)
A continuous method for the formation of cyclic carbonates from epoxides and carbon dioxide (CO2) is described. The catalysts used are inexpensive and effective in converting the reagents to the products in a residence time (t(R)) of 30 min. The
Bromine-catalyzed aziridination of olefins. A rare example of atom-transfer redox catalysis by a main group element.
Jeong JUK, et al.
Journal of the American Chemical Society, 120(27), 6844-6845 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service