Skip to Content
Merck
All Photos(2)

Key Documents

147214

Sigma-Aldrich

4-Amino-2-nitrophenol

97%

Synonym(s):

4-Hydroxy-3-nitroaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3(NO2)OH
CAS Number:
Molecular Weight:
154.12
Beilstein:
1368435
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

mp

125-127 °C (lit.)

functional group

nitro

SMILES string

Nc1ccc(O)c(c1)[N+]([O-])=O

InChI

1S/C6H6N2O3/c7-4-1-2-6(9)5(3-4)8(10)11/h1-3,9H,7H2

InChI key

WHODQVWERNSQEO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Amino-2-nitrophenol is the major metabolite of 2,4-dinitrophenol[1]. It is commonly used as semipermanent (nonoxidative) hair colorant and toner in permanent (oxidative) hair dye products[2].

Application

4-Amino-2-nitrophenol was used in determination of aminonitrophenols in hair dyes by differential pulse voltammetry and HPLC with electrochemical detection[3].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of Aminonitrophenols in Hair Dyes Using a Carbon Paste Electrode and a Boron-Doped Diamond Film Electrode-A Comparative Study.
Dejmkova H, et al.
International Journal of Electrochemical Science, 6, 3550-3563 (2011)
Hana Dejmkova et al.
Talanta, 85(5), 2594-2598 (2011-10-04)
Methods for determination of 2-amino-4-nitrophenol and 4-amino-2-nitrophenol, metabolites of 2,4-dinitrophenol, were developed using differential pulse (DP) voltammetry and HPLC with amperometric and spectrophotometric detection. The applicability of these methods was tested by the determination of the analytes in model samples
M A CAMERON
British journal of pharmacology and chemotherapy, 13(1), 25-29 (1958-03-01)
The effect of the mono- and di-nitrophenols and certain related compounds has been determined on the rate of oxygen consumption, the rate of carbon dioxide output and the rectal temperature of the Wistar albino rat.Of the compounds examined, only 2:4-dinitrophenol
M M Shahin et al.
Carcinogenesis, 3(7), 809-813 (1982-01-01)
Two samples of 4-amino-2-nitrophenol were tested for mutagenicity in Salmonella typhimurium strains TA1535, TA1537, TA1538, TA98, and TA100. Significant mutagenic activity was detected with a sample of this compound obtained from Aldrich (technical grade (Aldrich), TGA) both in the presence
Christina L Burnett et al.
International journal of toxicology, 28(6 Suppl 2), 217S-251S (2010-01-30)
2-Amino-3-nitrophenol, 2-amino-4-nitrophenol, 2-amino-5-nitrophenol, 4-amino-3-nitrophenol, 4-amino-2-nitrophenol, 2-amino-4-nitrophenol sulfate, 3-nitro-p-hydroxyethylaminophenol, and 4-hydroxypropylamino-3-nitrophenol are substituted aromatic compounds used as semipermanent (nonoxidative) hair colorants and as toners in permanent (oxidative) hair dye products. All ingredients in this group except 2-amino-4-nitrophenol sulfate, 2-amino-5-nitrophenol, and 4-amino-2-nitrophenol

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service