Skip to Content
Merck
All Photos(1)

Key Documents

W225002

Sigma-Aldrich

(−)-Carvyl acetate

mixture of cis and trans, ≥98%, FG

Synonym(s):

(1RS,5R)-5-Isopropenyl-2-methyl-2-cyclohexenyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18O2
CAS Number:
Molecular Weight:
194.27
FEMA Number:
2250
Beilstein:
2330666
EC Number:
Council of Europe no.:
2063
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.215
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

300
400

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

optical activity

[α]20/D -50°, neat

refractive index

n20/D 1.475 (lit.)

bp

77-79 °C/0.1 mmHg (lit.)

density

0.972 g/mL at 20 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; minty; sweet

SMILES string

CC(=C)C1CC=C(C)C(C1)OC(C)=O

InChI

1S/C12H18O2/c1-8(2)11-6-5-9(3)12(7-11)14-10(4)13/h5,11-12H,1,6-7H2,2-4H3

InChI key

YTHRBOFHFYZBRJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service