Skip to Content
Merck
All Photos(1)

Key Documents

L2807

Sigma-Aldrich

3,7-Dimethyl-1,6-octadien-3-yl acetate

97%

Synonym(s):

Linalyl acetate, 3,7-Dimethyl-1,6-octadien-3-yl acetate, Bergamol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2C(CH=CH2)(CH3)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
196.29
Beilstein:
1724500
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

6.8 (vs air)

vapor pressure

0.1 mmHg ( 20 °C)

Assay

97%

refractive index

n20/D 1.453 (lit.)

bp

220 °C (lit.)

density

0.901 g/mL at 25 °C (lit.)

SMILES string

C\C(C)=C\CCC(C)(OC(C)=O)C=C

InChI

1S/C12H20O2/c1-6-12(5,14-11(4)13)9-7-8-10(2)3/h6,8H,1,7,9H2,2-5H3

InChI key

UWKAYLJWKGQEPM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Sköld et al.
Contact dermatitis, 58(1), 9-14 (2007-12-25)
Fragrances are among the most common causes of allergic contact dermatitis. We have in previous studies shown that linalool, present in lavender oil, autoxidizes on air exposure, forming allergenic oxidation products. Oxidized linalool was found to be a frequent cause
E Barocelli et al.
Life sciences, 76(2), 213-223 (2004-11-03)
In this study the antinociceptive and the gastroprotective effects of orally administered or inhaled Lavandula hybrida Reverchon "Grosso" essential oil, and its principal constituents linalool and linalyl acetate were evaluated in rodents. Either when orally administered (100 mg/kg) or inhaled
Pio Maria Furneri et al.
European journal of medicinal chemistry, 52, 66-69 (2012-04-03)
Forty-two strains of Mycoplasma hominis (including PG21), 2 strain of Mycoplasma fermentans (Pg18 and K7), 1 strain of Mycoplasma pneumoniae (strain m129) were investigated for their susceptibilities to Citrus bergamia essential oil and to its major components (limonene, linalyl acetate
Simon Muhoho Njoroge et al.
Journal of agricultural and food chemistry, 51(14), 4029-4035 (2003-06-26)
The chemical changes and artifact formation in daidai (Citrus aurantium L. var. Cyathifera Y. Tanaka) cold-pressed peel oil upon storage at 20, 5, and -21 degrees C for 3, 6, and 12 months were investigated using capillary gas chromatography (GC)
Ali Reza Fakhari et al.
Journal of chromatography. A, 1098(1-2), 14-18 (2005-11-30)
A new method involving concurrent headspace solvent microextraction combined with continuous hydrodistillation (HD-HSME) for the extraction and pre-concentration of the essential oil of Lavandula angustifolia Mill. into a microdrop is developed. A microdrop of n-hexadecane containing n-heptadecane (as internal standard)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service