Skip to Content
Merck
All Photos(1)

Key Documents

13635

Sigma-Aldrich

Nickel(II) sulfate hexa-/ heptahydrate

DIN 50970 H, ≥20.6% Ni and Co basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NiSO4 · 6-7H2O
CAS Number:
Molecular Weight:
154.76 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder or crystals

grade

for analytical purposes

Agency

DIN 50970 H

form

powder or crystals

composition

Ni and Co, ≥20.6%

reaction suitability

reagent type: catalyst
core: nickel

impurities

≤0.05% insoluble in H2SO4

pH

≥3.5 (20 °C, 20%)

cation traces

As: ≤10 mg/kg
Cd: ≤50 mg/kg
Co: ≤5000 mg/kg
Cu: ≤20 mg/kg
Fe: ≤50 mg/kg
Pb: ≤20 mg/kg
Zn: ≤50 mg/kg

application(s)

battery manufacturing

SMILES string

[Ni++].[O-]S([O-])(=O)=O

InChI

1S/Ni.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

LGQLOGILCSXPEA-UHFFFAOYSA-L

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Curto et al.
Journal of the European Academy of Dermatology and Venereology : JEADV, 28(10), 1356-1362 (2014-03-13)
Topical corticosteroids (TC) consistently show effectiveness against itch, a paradigmatic symptom, in various eczemas. Rapid itch relief is a therapeutic goal. The early response of itch to TC has not been adequately studied. To assess the effect on itch of
Svetlana Paskaš et al.
Molecular carcinogenesis, 58(8), 1362-1375 (2019-04-19)
The main focus of this study is exploring the effect and mechanism of two HIV-protease inhibitors: Ritonavir and Ritonavir-nitric oxide (Ritonavir-NO) on in vitro growth of melanoma cell lines. NO modification significantly improved the antitumor potential of Ritonavir, as the
Fenxia Hou et al.
ALTEX, 37(3), 451-468 (2020-04-24)
There are currently no skin sensitization assays based on T cell activation. We built a novel in vitro model to assess T cell activation and test its performance to discriminate skin sensitizers from non-sensitizers using 52 reference chemicals. Jurkat Clone
Charlotte G Mortz et al.
Pediatric allergy and immunology : official publication of the European Society of Pediatric Allergy and Immunology, 24(1), 84-87 (2013-01-22)
We have previously reported patch test reactivity to nickel sulphate in a cohort of unselected infants tested repeatedly at 3-18 months of age. A reproducible positive reaction at 12 and 18 months was selected as a sign of nickel sensitivity provided a
Guangwei Si et al.
Lab on a chip, 12(7), 1389-1394 (2012-03-01)
We developed a multiple-channel microfluidic device for bacterial chemotaxis detection. Some characteristics such as easy operation, parallel sample adding design and fast result readout make this device convenient for most biology labs. The characteristic feature of the design is the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service