Skip to Content
Merck
All Photos(3)

Key Documents

F9252

Sigma-Aldrich

Folin & Ciocalteu′s phenol reagent

suitable for determination of total protein by Lowry method, 1.9-2.1 N

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12171500
NACRES:
NA.47

form

liquid

Quality Level

concentration

1.9-2.1 N

color

clear yellow
faint yellow to very dark yellow, and Faint Green-Yellow to Very Dark Green-Yellow

pH

<0.5 (20 °C)

density

1.240 g/cm3 at 20 °C

suitability

suitable for determination of total protein by Lowry method

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Folin & Ciocalteu′s phenol reagent is most commonly used in the Lowry method for determining protein concentration. It has also been used for the quantification of total phenolics. In this method, protein is pretreated with copper(II) in a modified biuret reagent (alkaline copper solution
stabilized with sodium potassium tartrate). Addition of the phenol reagent generates chromogens that give increasing absorbance between 550-750nm. Normally, absorbance at the peak (750nm) or shoulder (660nm) are used to quantitate protein concentrations between 1-100 mg/ml while absorbance at 550nm is used to quantitate higher protein concentrations.
In the absence of copper, color intensity is determined primarily by the tyrosine and tryptophan content of the protein, and to a lesser extent by cysteine and histidine. Copper(II) has no effect on color formation by tyrosine, tryptophan, or histidine, but reduces color formation due to cysteine.
Many modifications of the original assay procedure have been published, including methods for enhancing the color development, for determining the content of insoluble proteins, and for automating the procedure. Compounds including many buffers, chelating agents, detergents, and cyclic organic compounds can interfere with the Lowry protein assay.
Folin & Ciocalteu′s phenol reagent can also be used as a spray reagent in chromatographic procedures.

Application

Folin & Ciocalteu′s phenol reagent has been used for the estimation of total phenolic content in samples.

Other Notes

Folin & Ciocalteu′s phenol reagent does not contain phenol. Rather, the reagent will react with phenols and non-phenolic reducing substances to form chromogens that can be detected spectrophotometrically.

Linkage

This product is also available as part of a kit.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H J Fryer et al.
Analytical biochemistry, 153(2), 262-266 (1986-03-01)
The method of protein determination reported by Lowry et al. (1951, J. Biol. Chem. 193, 265-275) has been adapted for use with 96-well microtiter plates and an automatic microplate spectrophotometer. The spectrophotometer has been interfaced with a computer which plots
Picheswara Rao Polu et al.
BMC complementary and alternative medicine, 17(1), 457-457 (2017-09-13)
Tinospora cordifolia (Guduchi or Amrita) is an important drug of Ayurvedic System of Medicine and found mention in various classical texts for the treatment of diseases such as jaundice, fever, diabetes, cancer and skin disease etc. In view of its
Nutritional evaluation of some subtropical red and green seaweeds Part II. In vitro protein digestibility and amino acid profiles of protein concentrates.
Wong KH and Cheung PCK
Food Chemistry, 72, 11-17 (2001)
Lowry's method of protein estimation: some more insights.
S Sengupta et al.
The Journal of pharmacy and pharmacology, 45(1), 80-80 (1993-01-01)
David L Jones et al.
PloS one, 9(3), e90882-e90882 (2014-03-19)
Monitoring the properties of dissolved organic carbon (DOC) in soil water is frequently used to evaluate changes in soil quality and to explain shifts in freshwater ecosystem functioning. Using >700 individual soils (0-15 cm) collected from a 209,331 km(2) area

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service