Skip to Content
Merck
All Photos(1)

Documents

656895

Sigma-Aldrich

Nickel(II) sulfate

anhydrous, 99.99% trace metals basis

Synonym(s):

Nickel(2+) sulfate, Nickelous sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NiSO4
CAS Number:
Molecular Weight:
154.76
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

grade

anhydrous

Assay

99.99% trace metals basis

form

solid

impurities

≤150.0 ppm Trace Metal Analysis

application(s)

battery manufacturing

SMILES string

[Ni++].[O-]S([O-])(=O)=O

InChI

1S/Ni.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

LGQLOGILCSXPEA-UHFFFAOYSA-L

General description

Nickel(II) sulfate is an inorganic compound that exhibits high solubility in water. It is commonly used in electroplating, as a chemical intermediate in the preparation of other nickel salts, and as a source of nickel ions for various processes. Additionally, it has applications in the battery industry and as a catalyst in organic synthesis.
Properties of NiSO4:      
  • Highly soluble in water     
  • It can participate in redox reactions  
  • It can form complexes with various ligands

Application

Nickel(II) sulfate can be used as a precursor to synthesize:    
  • Nickel-based electrocatalysts for oxygen evolution.      
  • Ni-doped titanium dioxide nanoparticles for the photocatalytic degradation of organic pollutants such as methyl orange.     
  • Spherical silica particles coated with Ni, which find application in the field of ceramic industry and in catalysis.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 1 - Carc. 1A Inhalation - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

respiratory tract irritation

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Curto et al.
Journal of the European Academy of Dermatology and Venereology : JEADV, 28(10), 1356-1362 (2014-03-13)
Topical corticosteroids (TC) consistently show effectiveness against itch, a paradigmatic symptom, in various eczemas. Rapid itch relief is a therapeutic goal. The early response of itch to TC has not been adequately studied. To assess the effect on itch of
Michela Notarangelo et al.
EBioMedicine, 43, 114-126 (2019-05-03)
Extracellular vesicles (EVs) are secreted membranous particles intensively studied for their potential cargo of diagnostic markers. Efficient and cost-effective isolation methods need to be established for the reproducible and high-throughput study of EVs in the clinical practice. We designed the
Xiaohua Xu et al.
Particle and fibre toxicology, 9, 40-40 (2012-11-07)
It has been well recognized that toxicity of fine ambient air particulate matter (PM(2.5)) may depend on its chemical constituents, including components such as soluble metals that may theoretically exert distinctive effects. We have recently demonstrated an important effect of
K Amudha et al.
Chemico-biological interactions, 193(1), 57-64 (2011-05-24)
This study was conducted to investigate the beneficial role of naringin on nickel induced nephrotoxicity. Nickel (Ni) (20mg/kg body weight (b.w.) was administered intraperitoneally (i.p.) for 20 days. Naringin was administered orally (20, 40 and 80 mg/kg b.w.) with i.p.
John C Steele et al.
Dermatitis : contact, atopic, occupational, drug, 23(2), 61-70 (2012-06-02)
Patients with a sore or burning mouth associated with clinically normal oral mucosa present a difficult diagnostic challenge. The objective of this study was to assess the value of patch testing in patients with burning mouth syndrome. We retrospectively reviewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service