Skip to Content
Merck
All Photos(4)

Key Documents

654507

Sigma-Aldrich

Nickel(II) chloride hexahydrate

99.9% trace metals basis

Synonym(s):

Nickel chloride hexahydrate 2, Nickel dichloride hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NiCl2 · 6H2O
CAS Number:
Molecular Weight:
237.69
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
solid
Pricing and availability is not currently available.

grade

for analytical purposes

Assay

99.9% trace metals basis

form

solid

impurities

≤1500.0 ppm Trace Metal Analysis

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].Cl[Ni]Cl

InChI

1S/2ClH.Ni.6H2O/h2*1H;;6*1H2/q;;+2;;;;;;/p-2

InChI key

LAIZPRYFQUWUBN-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Nickel(II) chloride hexahydrate is a water-soluble inorganic compound. It is a cost-effective compound that finds applications in electroplating, nickel-plating, and as an NH3 absorbent in gas masks.

Application

Nickel(II) chloride hexahydrate can be used as:      
  • A precursor for nickel oxide (NiO) films, which are used as hole transport layers in dye-sensitized solar cells (DSSCs).     
  • An additive to the electron transport layer (ETL) in perovskite solar cells to improve their performance, particularly the open-circuit voltage.
  • An effective dopant for synthesizing conductive polymer composites for potential applications in flexible electronics, and sensors.     
  • A precursor in the synthesis of nickel nanothorn particles via hydrothermal method.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

NiCl2.6H2O: an efficient catalyst precursor for phosphine-free Heck and Sonogashira cross-coupling reactions
Nowrouzi N and Zarei M
Tetrahedron, 71(41), 7847-7852 (2015)
Kariana Moreno-Sader et al.
ACS omega, 4(6), 10834-10844 (2019-08-29)
Nanocomposites composed of polyacrylamide and nanoclay were synthesized via free-radical cross-linking polymerization and used to adsorb Co2+ and Ni2+ ions from water. The polyacrylamide (PAM)/sodium montmorillonite (Na-MMT) nanocomposites were characterized by Fourier transform infrared spectroscopy, X-ray diffraction, and scanning electron
Synthesis of tetra-substituted pyrroles, a potential phosphodiesterase 4B inhibitor, through nickel (II) chloride hexahydrate catalyzed one-pot four-component reaction.
Khan AT, et al.
Tetrahedron Letters, 53(32), 4145-4150 (2012)
Jenilee Way et al.
Nuclear medicine and biology, 40(3), 430-436 (2013-01-12)
4-[(18)F]Fluorobenzylamine ([(18)F]FBA) is an important building block for the synthesis of (18)F-labeled compounds. Synthesis of [(18)F]FBA usually involves application of strong reducing agents like LiAlH4 which is challenging to handle in automated synthesis units (ASUs). Therefore, alternative methods for the
R Prabhakaran et al.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry, 18(2), 233-247 (2013-01-01)
Three new nickel(II) thiosemicarbazone complexes have been synthesized and characterized by analytical, spectral, and single-crystal X-ray diffraction studies. In complex 1, the ligand 2-hydroxy-1-naphthaldehydethiosemicarbazone coordinated as a monobasic tridentate donor, whereas in complexes 2 and 3, the ligands salicylaldehyde-4(N)-ethylthiosemicarbazone and

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service