Skip to Content
Merck
All Photos(2)

Documents

229598

Sigma-Aldrich

Cobalt(II) sulfate hydrate

99.998% trace metals basis

Synonym(s):

Cobaltous sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CoSO4 · xH2O
CAS Number:
Molecular Weight:
155.00 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.998% trace metals basis

form

crystals and lumps

impurities

≤25.0 ppm Trace Rare Earth Analysis

density

2.03 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

O.[Co++].[O-]S([O-])(=O)=O

InChI

1S/Co.H2O4S.H2O/c;1-5(2,3)4;/h;(H2,1,2,3,4);1H2/q+2;;/p-2

InChI key

BGORGFZEVHFAQU-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Cobalt(II) sulfate hydrate (CoSO4.H2O) can be used in the synthesis of coordination polymers. It can also be used as a starting material in the synthesis of cobalt sulfides hexagonal nanocrystallites.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michal Nowak et al.
Luminescence : the journal of biological and chemical luminescence, 34(2), 183-192 (2019-01-25)
Oxidative reactions can result in the formation of electronically excited species that undergo radiative decay depending on electronic transition from the excited state to the ground state with subsequent ultra-weak photon emission (UPE). We investigated the UPE from the Fe2+
Copper (II) and cobalt (II) coordination polymers with bridging 1, 2, 4, 5-benzenetetracarboxylate and N-methylimidazole: coordination number-determined sheet topology
Cheng D, et al.
Journal of the Chemical Society, Part 1, 4555-4560 (2002)
In situ deposition of Co9S8 nanocrystallite on its single-crystal flakes at low temperatures
Zhang Y, et al.
Bulletin of the Chemical Society of Japan, 78(2), 277-280 (2005)
Huisu Lee et al.
Fish & shellfish immunology, 111, 119-126 (2021-01-28)
There are few reports of dietary Cu (copper) toxicity to Korean bullhead, Pseudobagrus fulvidraco, and little is known about recovery from dietary Cu exposure. In this study, P. fulvidraco (mean length 16.9 ± 1.38 cm, and mean weight 53.2 ± 1.22 g) were exposed for 4

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service