Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

710873

Sigma-Aldrich

Methyllithium solution

greener alternative

3% in 2-Methyltetrahydrofuran/cumene

Synonym(s):

Lithium methanide, MeLi

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH3Li
CAS Number:
Molecular Weight:
21.98
Beilstein/REAXYS Number:
3587162
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

form

liquid

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

concentration

3% in 2-Methyltetrahydrofuran/cumene

density

0.860 g/mL at 25 °C

greener alternative category

storage temp.

2-8°C

SMILES string

[Li]C

InChI

1S/CH3.Li/h1H3;

InChI key

DVSDBMFJEQPWNO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 1B - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 1

target_organs

Respiratory system

supp_hazards

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 2

flash_point_f

19.4 °F

flash_point_c

-7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erik Selander et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(20), 6395-6400 (2015-04-29)
Interactions among microscopic planktonic organisms underpin the functioning of open ocean ecosystems. With few exceptions, these organisms lack advanced eyes and thus rely largely on chemical sensing to perceive their surroundings. However, few of the signaling molecules involved in interactions

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service