Skip to Content
MilliporeSigma

Skip To

T7771

Ethyl 2-mercaptoacetate

Synonym(s):

Thioglycolic acid ethyl ester

Sign In to View Organizational & Contract Pricing.

About This Item

EC Number:
210-800-3
UNSPSC Code:
12352200


density

1.1 g/mL at 25 °C (lit.)

storage temp.

−20°C





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library