Skip to Content
MilliporeSigma

Skip To

ABC60

Anti-Endostatin Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Collagen alpha-1(XVIII) chain, Endostatin

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
1


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human, mouse

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... COL18A1(80781)
mouse ... Col18A1(12822)

General description

Endostatin is a proteolytic cleavage product from the C-terminus of Collagen XVIII, which is itself a proteoglycan/collagen found in blood vessel walls and in basement membranes. Endostatin is a compact globular fold-containing protein with a Zn2+ binding site near its amino terminus. Originally identified in conditioned media of murine hemangioendothelioma (EOMA) cells, endostatin is generated from the parent collagen XVIII molecule involving intermediate glycosylated forms and the proteolytic activity of proteases such as cathepsin L, metalloproteases, and elastase. Endostatin has a wide-spread tissue expression and has been shown to co-localize with fibulins, nidogen-2 in basement membranes, and elastic fibers of veins. Endostatin has also been shown to cause a G1 arrest in endothelial cells and to induce apoptosis.
~28 kDa observed.
A ~28 kDa endostatin fragment has been observed in western blot as a result of MMP-7 cleavage of collagen XVIII (Chang, J., et al. (2003). Invest Ophthalmol Vis Sci. 44:832).

Immunogen

Epitope: Nonhelical Region 11 Domain
Linear peptide corresponding to the nonhelical region 11 of mouse Endostatin.

Application

Anti-Endostatin Antibody detects level of Endostatin & has been published & validated for use in WB, IH(P).
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected Endostatin in mouse lung tissue.
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)

Biochem/physiol Actions

This antibody recognizes Endostatin at the nonhelical region 11 domain.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
A431 cell lysate
Evaluated by Western Blot in A431 cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected Endostatin in 10 µg of A431 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Replaces: AB1880

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB4503213SAB4503212SAB4500395
conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

Gene Information

human ... COL18A1(80781)
mouse ... Col18A1(12822)

Gene Information

human ... COL18A1(80781)

Gene Information

human ... COL18A1(80781)

Gene Information

human ... COL12A1(1303)

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

technique(s)

ELISA: 1:5000, immunofluorescence: 1:100-1:500, immunohistochemistry: 1:50-1:100

technique(s)

ELISA: 1:20000, immunohistochemistry: 1:50-1:100

technique(s)

ELISA: 1:1000, immunofluorescence: 1:100-1:500, immunohistochemistry: 1:50-1:100


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Rui Tang et al.
The Journal of biological chemistry, 291(52), 26913-26921 (2016-11-09)
Vascular smooth muscle cells (SMCs) and endothelial cells (ECs) are in close contact with blood vessels. SMC phenotypes can be altered during pathological vascular remodeling. However, how SMC phenotypes affect EC properties remains largely unknown. In this study, we found



Global Trade Item Number

SKUGTIN
ABC6004053252640681

Questions

Reviews

No rating value

Active Filters