Skip to Content
MilliporeSigma

Skip To

391069

Trichlorocyclohexylsilane

98%

Synonym(s):

Cyclohexyltrichlorosilane

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C6H11Cl3Si
CAS Number:
Molecular Weight:
217.60
EC Number:
202-639-2
UNSPSC Code:
12352101
PubChem Substance ID:
Beilstein/REAXYS Number:
2935257
MDL number:
Assay:
98%


assay

98%

refractive index

n20/D 1.478 (lit.)

bp

90 °C/10 mmHg (lit.)

density

1.232 g/mL at 25 °C (lit.)

SMILES string

Cl[Si](Cl)(Cl)C1CCCCC1

InChI

1S/C6H11Cl3Si/c7-10(8,9)6-4-2-1-3-5-6/h6H,1-5H2

InChI key

SIPHWXREAZVVNS-UHFFFAOYSA-N



pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk

WGK 3

flash_point_f

201.2 °F - closed cup

flash_point_c

94 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters