Recommended Products
assay
99%
form
solid
mp
53-55 °C (lit.)
SMILES string
CC(=O)C12C[C@H]3C[C@H](C[C@H](C3)C1)C2
InChI
1S/C12H18O/c1-8(13)12-5-9-2-10(6-12)4-11(3-9)7-12/h9-11H,2-7H2,1H3/t9-,10+,11-,12-
InChI key
DACIGVIOAFXPHW-CDECOKDKSA-N
General description
1-Adamantyl methyl ketone, a sterically bulky ketone substrate, was reduced to the corresponding alcohol with excellent optical purity.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
226.4 °F - closed cup
flash_point_c
108.00 °C - closed cup
ppe
Eyeshields, Gloves, type N95 (US)
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Organic & biomolecular chemistry, 4(14), 2690-2695 (2006-07-11)
In our effort to search for effective carbonyl reductases, the activity and enantioselectivity of a carbonyl reductase from Sporobolomyces salmonicolor have been evaluated toward the reduction of a variety of ketones. This carbonyl reductase (SSCR) reduces a broad spectrum of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service