HPLC Analysis of Immunoglobulins (IgM) on Q Sepharose™ Fast Flow
![HPLC Analysis of Immunoglobulins (IgM) on Q Sepharose™ Fast Flow application for HPLC](/deepweb/assets/sigmaaldrich/product/images/214/173/94e8a518-12b9-45f0-9c0a-74e4609b2010/800/94e8a518-12b9-45f0-9c0a-74e4609b2010.jpg)
材料
分析柱
產品號碼
描述
訂價
CONDITIONS
media
Q Sepharose Fast Flow (Q1126-100ML)
column
30 cm × 50 mm I.D. (Q1126-100ML)
mobile phase
[A] 0.02 M monosodium phosphate, pH 6.5; [B] A + 0.5 M sodium chloride, pH 6.5
flow rate
25 cm/hr
detector
UV, 280 nm
injection
cell culture supernatant
描述
法律資訊
Sepharose is a trademark of Cytiva